Skip to Content
Merck
All Photos(7)

Key Documents

D6944

Sigma-Aldrich

Anti-Dopamine Transporter (N-terminal) antibody produced in rabbit

enhanced validation

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-DAT, Anti-DAT1, Anti-SLC6A3, Anti-Solute carrier family 6, member 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~50 kDa

species reactivity

mouse (predicted), rat, human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~1.0 mg/mL

technique(s)

immunofluorescence: 5-10 μg/mL using SH-SY-5Y cells
western blot: 0.5-1 μg/mL using rat brain tissue extract

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC6A3(6531)
mouse ... Slc6a3(13162)
rat ... Slc6a3(24898)

General description

Dopamine transporter belongs to a family of biogenic amine transporters that include the norepinephrine, serotonin (SERT), and vesicular monoamine transporters. These transporters are members of a larger Na+/Cl--dependent neurotransmitter transporter family.
Solute carrier family 6 member 3 (SLC6A3) is mapped to human chromosome 5p15.33. It is also known as the dopamine transporter (DAT) and has a zinc binding site and 12 transmembrane region.

Immunogen

synthetic peptide corresponding to amino acids 42-59 of rat dopamine transporter conjugated to KLH. This sequence is highly conserved in mouse DAT (single amino acid substitution) and has lower homology (72% identity) to human DAT.

Application

Anti-Dopamine Transporter antibody produced in rabbit is suitable for western blot analysis at a working concentration of 0.5-1μg/mL using rat brain extract (cytosolic S1 fraction) and HEK-293T cells expressing human DAT1. It was used as a primary antibody in western analysis for dopamine transporter in several studies.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunocytochemistry (1 paper)
Immunohistochemistry (1 paper)
Western Blotting (1 paper)

Biochem/physiol Actions

DAT regulates dopaminergic-neurotransmission. It is localized on presynaptic nerve terminals and mediates the active reuptake of dopamine from the synapse, thus terminating the neurotransmitter signal. It is the primary site for the action of neurotoxins, such as MPTP, and the drugs of abuse, cocaine and amphetamine. It has been implicated in a number of dopamine-related disorders, including attention deficit hyperactivity disorder (ADHD), bipolar disorder, clinical depression, and alcoholism. DAT may also have a role in selective vulnerability of nigrostriatal dopaminergic neurons in Parkinson′s disease as it can transport dopamine neurotoxins.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Enrique Garea-Rodríguez et al.
PloS one, 11(2), e0149776-e0149776 (2016-02-24)
Cerebral dopamine neurotrophic factor (CDNF) belongs to a newly discovered family of evolutionarily conserved neurotrophic factors. We demonstrate for the first time a therapeutic effect of CDNF in a unilateral 6-hydroxydopamine (6-OHDA) lesion model of Parkinson's disease in marmoset monkeys.
A Storch et al.
Journal of neural transmission (Vienna, Austria : 1996), 111(10-11), 1267-1286 (2004-10-14)
The carrier molecule that transports dopamine (DA) into dopamine neurons by an electrogenic, Na(+)- and Cl(-)-transport-coupled mechanism is known as the dopamine transporter (DAT). This uptake system is exclusively expressed in DA neurons with significantly higher levels of DAT expression
Kristopher M Kahlig et al.
The Journal of biological chemistry, 279(10), 8966-8975 (2003-12-31)
Dopaminergic neurotransmission is fine-tuned by the rate of removal of dopamine (DA) from the extracellular space via the Na(+)/Cl(-)-dependent DA transporter (DAT). DAT is a target of psychostimulants such as amphetamine (AMPH) and cocaine. Previously, we reported that AMPH redistributes
Symmetrical dimer of the human dopamine transporter revealed by cross-linking Cys-306 at the extracellular end of the sixth transmembrane segment.
Hastrup H, et al.
Proceedings of the National Academy of Sciences of the USA, 98(18), 10055-10060 (2001)
Takayuki Nakagawa et al.
PloS one, 6(9), e24865-e24865 (2011-10-08)
Repeated intermittent exposure to psychostimulants and morphine leads to progressive augmentation of its locomotor activating effects in rodents. Accumulating evidence suggests the critical involvement of the mesocorticolimbic dopaminergic neurons, which project from the ventral tegmental area to the nucleus accumbens

Articles

Protein-based drug transporters are expressed in Sf9 cells. Understanding the specific mechanisms of tumor cell transporters is an essential aspect of chemotherapeutic drug design.

Protein-based drug transporters are expressed in Sf9 cells. Understanding the specific mechanisms of tumor cell transporters is an essential aspect of chemotherapeutic drug design.

Protein-based drug transporters are expressed in Sf9 cells. Understanding the specific mechanisms of tumor cell transporters is an essential aspect of chemotherapeutic drug design.

Protein-based drug transporters are expressed in Sf9 cells. Understanding the specific mechanisms of tumor cell transporters is an essential aspect of chemotherapeutic drug design.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service