Skip to Content
Merck
All Photos(1)

Key Documents

AB1591P

Sigma-Aldrich

Anti-Dopamine Transporter Antibody

Chemicon®, from rabbit

Synonym(s):

DAT

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... SLC6A3(6531)

General description

Plasmalemmal neurotransmitter transporters sequester synaptic and peri synaptic transmitter into presynaptic elements. The Dopamine Transporter (DAT) is responsible for the reaccumulation of dopamine after it has been released. Levels of DAT protein expression are altered by chronic drug administration. The activity of the DAT reuptake carrier is sodium dependent, and it is suspected to play a role in such neurologic and psychiatric disorders as Parkinson′s disease, Tourette′s disease, schizophrenia, and addiction. It is a 12 transmembrane domain transporter with the N and C terminal regions located within the cytoplasm.

Specificity

Recognizes Dopamine transporter (DAT). By Western blot the antibody recognizes a broad 80 kDa band in rat striatum (Lew et al. 1992; Freed et al. 1995; Vaughan et al. 1996) and not cerebellum. Rat DAT contains various carbohydrate moieties and neuraminidase and N-Glycanase treatment reduces its molecular weight to about 55 kDa.

Immunogen

An 18 amino acid peptide sequence near the NH2-terminus of rat brain dopamine transporter coupled to KLH (Kilty et al. 1991; Giros et al. 1991; Shimada et al. 1991; Kitayma 1992). This region is presumably the cytoplasmic domain.

Application

Detect Dopamine Transporter using this Anti-Dopamine Transporter Antibody validated for use in ELISA, IH, IP & WB.
Western blot: 1-10 μg/mL using ECL. An antibody raised against a similar peptide has been shown to react with a broad 75-80 kDa band in rat striatum (Hastrup et al., 2001) and not cerebellum. Rat DAT contains various carbohydrate moieties and neuraminidase and N-Glycanase treatment reduces its molecular weight to about 55 kDa.

Immunoprecipitation: 2-10 μg of antibody (Chang et al., 2001)

Immunohistochemistry: 1:250-1:500 on frozen sections of tissues fixed with 4% paraformaldehyde (Chang et al., 2001; Ourednik, et al., 2002). 0.05% trypsin for 10-15min has been found effective in exposing the epitope in some preparations.

ELISA: 0.5-1.0 μg/mL using 1 μg/mL of DAT control peptide (Catalog Number AG296) per well.

Optimal working dilutions must be determined by end user.

Target description

80 kDa

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silke Frahm et al.
Behavioural brain research, 339, 153-168 (2017-11-29)
Alpha-Synuclein (α-Syn) accumulation is considered a major risk factor for the development of synucleinopathies such as Parkinson's disease (PD) and dementia with Lewy bodies. We have generated mice overexpressing full-length human α-Syn fused to a membrane-targeting signal sequence under the
Jian Huang et al.
Brain and behavior, 10(2), e01533-e01533 (2020-01-17)
This study aims to establish a methamphetamine (METH)-induced behavioral sensitization model using tree shrews, as well as to measure the protein expression of the dopamine D3 receptor (D3R) and dopamine transporter (DAT). Forty tree shrews were equally and randomly divided
Michael Y Aksenov et al.
Neurotoxicology, 29(6), 971-977 (2008-07-09)
Drug abuse is a risk factor for neurological complications in HIV infection. Cocaine has been shown to exacerbate HIV-associated brain pathology and enhance neurotoxicity of HIV-1 Tat and gp120 proteins. In this study, we found that the selective inhibitor of
Kirill Shumilov et al.
PloS one, 13(8), e0203135-e0203135 (2018-08-30)
The striatum is a complex structure in which the organization in two compartments (striosomes and matrix) have been defined by their neurochemical profile and their input-output connections. The striosomes receive afferences from the limbic brain areas and send projections to
Mauro Ceccanti et al.
Addiction biology, 21(4), 776-787 (2015-05-06)
Ethanol (EtOH) exposure during pregnancy induces cognitive and physiological deficits in the offspring. However, the role of paternal alcohol exposure (PAE) on offspring EtOH sensitivity and neurotrophins has not received much attention. The present study examined whether PAE may disrupt

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service