Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Dulbecco’s Phosphate Buffered Saline
Synonym(s):
DPBS
Products
Product Category
buffers (11)
Brand
SAFC (4)
Sigma-Aldrich (5)
Physical Form
Glucose (g/L)
Sodium Pyruvate (g/L)
Quality Segment
Sterilization Method
Technique
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
D8537
Dulbecco’s Phosphate Buffered Saline,
liquid, Modified, without calcium chloride and magnesium chloride, liquid, sterile-filtered, suitable for cell culture
Expand
View Pricing
D1408
Dulbecco’s Phosphate Buffered Saline,
Liquid (10X), without MgCl₂ and CaCl₂, sterile-filtered, suitable for cell culture
Expand
View Pricing
D8662
Dulbecco’s Phosphate Buffered Saline,
Liquid, with MgCl₂ and CaCl₂, sterile-filtered, suitable for cell culture
Expand
View Pricing
D5652
Dulbecco’s Phosphate Buffered Saline,
Modified, without calcium chloride and magnesium chloride, powder, suitable for cell culture
Expand
View Pricing
59331C
Dulbecco’s Phosphate Buffered Saline,
liquid, sterile-filtered, DPBS Modified 10X, without calcium, without magnesium, suitable for cell culture
Expand
View Pricing
TMS-012
Dulbecco’s Phosphate Buffered Saline,
1X, liquid, w/o Ca++ & Mg++, Endotoxin-Free
Expand
View Pricing
D1283
Dulbecco’s Phosphate Buffered Saline,
With calcium chloride and magnesium chloride, 10×, liquid, sterile-filtered, suitable for cell culture
Expand
View Pricing
59321C
Dulbecco’s Phosphate Buffered Saline,
liquid, sterile-filtered, DPBS Modified, without calcium, without magnesium, suitable for cell culture
Expand
View Pricing
D5773
Dulbecco’s Phosphate Buffered Saline,
Without calcium chloride, powder, suitable for cell culture
Expand
View Pricing
56064C
Dulbecco’s Phosphate Buffered Saline,
dry powder, DPBS Modified, without calcium, without magnesium, suitable for cell culture
Expand
View Pricing
59300C
Dulbecco’s Phosphate Buffered Saline,
liquid, sterile-filtered, with calcium, with magnesium, suitable for cell culture
Expand
View Pricing
D4031
Dulbecco’s Phosphate Buffered Saline,
liquid, Modified, with 36 mg sodium pyruvate, 50 mg streptomycin sulfate, 100 mg kanamycin monosulfate, 1000 mg glucose/L and CaCl₂, liquid, 0.1 μm filtered, suitable for cell culture
Expand
View Pricing