D4879
N,N′-Bis(hydroxymethyl)urea
≥95%
Synonym(s):
1,3-Bis(hydroxymethyl)urea, DMU, Dimethylolurea
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
HOCH2NHCONHCH2OH
CAS Number:
Molecular Weight:
120.11
Beilstein:
1811870
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
≥95%
form
powder
mp
125 °C (dec.) (lit.)
storage temp.
2-8°C
SMILES string
OCNC(=O)NCO
InChI
1S/C3H8N2O3/c6-1-4-3(8)5-2-7/h6-7H,1-2H2,(H2,4,5,8)
InChI key
QUBQYFYWUJJAAK-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Syyed Adnan Raheel Shah et al.
Journal of safety research, 69, 201-215 (2019-06-27)
An efficient decision-making process is one of the major necessities of road safety performance analysis for human safety and budget allocation procedure. During the road safety analysis procedure, data envelopment analysis (DEA) supports policymakers in differentiating between risky and safe
Wasim I M Sultan et al.
Conflict and health, 12, 44-44 (2018-11-10)
The structure, function, and capacity of the health care system in the Occupied Palestinian Territories (OPT) had been largely shaped by the complex political history of the country. Since the establishment of the Palestinian Authority in 1994, the reform efforts
Yuanyuan Wang et al.
PloS one, 15(12), e0243282-e0243282 (2020-12-04)
There are concerns that lumbar spine imaging represents low value care. Our aim was to examine the use of lumbar spine imaging [radiography, computed tomography (CT), magnetic resonance imaging (MRI)] over 20 years, and costs and person-level characteristics of imaging
Laura de Almeida Botega et al.
Health care management science, 23(3), 443-452 (2020-05-07)
This paper assesses the economic efficiency of Brazilian general hospitals that provide inpatient care for the Unified Health System (SUS). We combined data envelopment analysis (DEA) and spatial analysis to identify predominant clusters, measure hospital inefficiency and analyze the spatial
Angeliki Flokou et al.
Health care management science, 20(4), 467-484 (2016-04-14)
This paper evaluates the technical efficiency of 71 Greek public hospitals and examines potential efficiency gains from 13 candidate mergers among them. Efficiency assessments are performed using bootstrapped Data Envelopment Analysis (DEA) whilst merger analysis is conducted by applying the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service