Skip to Content
Merck
All Photos(3)

Key Documents

M86804

Sigma-Aldrich

N-Methylurea

97%

Synonym(s):

1-Methylurea, N-Methylurea

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3NHCONH2
CAS Number:
Molecular Weight:
74.08
Beilstein:
878189
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

form

crystals

SMILES string

CNC(N)=O

InChI

1S/C2H6N2O/c1-4-2(3)5/h1H3,(H3,3,4,5)

InChI key

XGEGHDBEHXKFPX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V V Matveev
Tsitologiia, 31(12), 1459-1465 (1989-12-01)
Following short-term local applications of high doses of N-methyl-N-nitrozourea and 7,2-dimethylbenz(a)antracen, the intercellular adhesion in the distal colon epithelium of ice retains increased at least for a month. The magnitude of this shift and its duration ops both with moving
P J Groenen et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 26(3), 215-225 (1988-03-01)
N-Nitroso compounds may well rank high among the genotoxic carcinogens present in our environment. Small amounts of such compounds may be formed in the human stomach after consumption of high-nitrate vegetables. Volatile nitrosamines can be conveniently determined but reliable methods
A J Streeter et al.
Drug metabolism and disposition: the biological fate of chemicals, 18(4), 447-452 (1990-07-01)
The single-dose toxicokinetics of monomethylamine has been characterized in the rat by HPLC assay of serial blood samples. Biphasic first-order elimination was observed following an iv bolus dose of 19 mumol/kg with a terminal half-life of 19.1 +/- 1.3 min
J Guzmán Rincón et al.
Mutation research, 412(1), 69-81 (1998-03-21)
The in vivo nitrosation capacity of third-instar larvae of Drosophila melanogaster was assessed using the wing somatic mutation and recombination test (SMART). Larvate derived from two different crosses, the standard cross (ST) and the high bioactivation cross (HB) both involving
Nuzhat Gull et al.
Colloids and surfaces. B, Biointerfaces, 51(1), 10-15 (2006-06-30)
The present study highlights the fact that the effect of additives (urea, monomethylurea, thiourea) on the supramolecular assemblies and proteins is strikingly similar. To investigate the effect, a viscometeric study on sphere-to-rod transition (s-->r) was undertaken in a system (3.5%

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service