Skip to Content
Merck
All Photos(1)

Key Documents

D4154

Sigma-Aldrich

Plasmid DNA from E. coli RRI

pUC18, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41105600
NACRES:
NA.85
Pricing and availability is not currently available.

biological source

Escherichia coli (RRI)

Quality Level

grade

for molecular biology

form

buffered aqueous solution

shipped in

dry ice

storage temp.

−20°C

SMILES string

[P](=O)(OC2C(OC(C2)N)CO[P](=O)(OC3C(OC(C3)N)CO)O)(OCC1OC(CC1O)N)O

InChI

1S/C15H31N3O13P2/c16-13-1-7(20)11(28-13)5-25-32(21,22)31-9-3-15(18)29-12(9)6-26-33(23,24)30-8-2-14(17)27-10(8)4-19/h7-15,19-20H,1-6,16-18H2,(H,21,22)(H,23,24)

InChI key

AWBASQCACWFTGD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Plasmid DNA from Escherichia coli RRI confers ampicillin resistance and complement defects in β-galactosidase in appropriate host strains. The multiple cloning site (MCS) is within the β-galactosidase gene. pUC8 and 9 have nine unique sites within the MCS while pUC18 and 19 have thirteen.

Application

Plasmid DNA from Escherichia coli RRI has been used in polymerase chain reaction (PCR).[1]
Plasmid DNA from E. coli RRI has also been used for in vitro DNA transfection studies.[2]

Biochem/physiol Actions

These plasmids confer ampicillin resistance and complement defects in β-galactosidase in appropriate host strains. The multiple cloning site (MCS) is within the β-galactosidase gene; pUC8 and 9 have nine unique sites within the MCS while pUC18 and 19 have thirteen.
Foreign DNA inserted at the MCS abolishes the ability to catabolize lactose. Lactose-positive, ampicillin-resistant colonies (host strain containing plasmid) form blue colonies on plates containing ampicillin and X-Gal; lactose-negative, ampicillin-resistant colonies (host strain containing plasmid with foreign DNA inserted at the MCS) form white colonies on this medium. The orientations of the MCS regions in the pUC plasmids are analogous to those of the corresponding M13 phage.

Components

pUC18 Plasmid DNA, 10 μg is supplied at approximately 0.5 mg/ml in 10 mM Tris-HCl (pH 8.0) with 1 mM EDTA.

Principle

Foreign DNA inserted at the MCS interrupts the β-galactosidase gene and abolishes the ability to catabolize lactose. Lactose-positive, ampicillin-resistant colonies (host strain containing plasmid) form blue colonies on plates containing ampicillin and X-Gal; lactose-negative, ampicillin-resistant colonies (host strain containing plasmid with foreign DNA inserted at the MCS) form white colonies on this medium.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Nucleoside phosphocholine amphiphile for in vitro DNA transfection
Moreau L, et al.
Molecular Biosystems, 1(3), 260-264 (2005)
Quantification of beta-human papillomavirus DNA by real-time PCR.
Weissenborn SJ
Nature Protocols, 5(1), 1-13 (2010)
Asife Sahinarslan et al.
Acta cardiologica, 68(3), 291-297 (2013-07-26)
Both computed tomography (CTA) and conventional angiography (CCA) can provide direct visualization of the coronary arteries. The aim of the present study was to compare the radiation exposure between CTA and CCA and to search whether this amount of radiation
M Iu Mazina et al.
Tsitologiia, 55(4), 218-224 (2013-07-24)
DNA replication begins from multiple sites distributed thoughout the genome and named replication origins. Despite the increasing amount of data on the properties of replication origins, it is still unknown what factors(s) is the primary determinant of ORC localization. Su(Hw)
Jong Bum Lee et al.
Journal of biomedical nanotechnology, 9(7), 1245-1249 (2013-08-06)
Recently, DNA has been used to guide the self-assembly of functional materials. Based on programmability of DNA, branched DNA nanostructures were created and precisely labeled with quantum dots and gold nanoparticles. The precise molecular recognition of DNA allows the precise

Questions

1–2 of 2 Questions  
  1. What is the sequence of plasmid pUC18 (product number D4154) and the number of copies of the plasmid contained in this product? It will be used as an internal control for PCR amplification.

    What is the sequence of plasmid pUC18 (product number D4154) and the number of copies of the plasmid contained in this product? It will be used as an internal control for PCR amplification.

    1 answer
    1. The GenBank accession number for the sequence of the high-copy number plasmid pUC18 is VB0025.

      The following link contains the sequence for this plasmid: https://www.addgene.org/50004/sequences/#depositor-full.

      The plasmid is 2,686 base pairs in length and the molecular weight is approximately 1.75 × 10⁶ daltons, so the 10µg should be 3.4 x 10^12 copies.

      Helpful?

  2. user manual for Puc18?

    1 answer
    1. The Puc18 is not supplied with a user manual or specific protocol. Please see the link below to the product data sheet which provides additional information on this plasmid: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/263/858/d4154dat.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service