Skip to Content
Merck
All Photos(1)

Documents

D2631

Sigma-Aldrich

Dihexadecyl phosphate

Synonym(s):

DHP, Dicetyl phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)15O]2P(O)OH
CAS Number:
Molecular Weight:
546.85
Beilstein:
1717117
EC Number:
MDL number:
UNSPSC Code:
51191904
PubChem Substance ID:
NACRES:
NA.25

description

anionic

form

powder

mol wt

546.85 g/mol

technique(s)

DNA extraction: suitable

mp

74-75 °C (lit.)

lipid type

phospholipids

storage temp.

−20°C

SMILES string

OP(OCCCCCCCCCCCCCCCC)(OCCCCCCCCCCCCCCCC)=O

InChI

1S/C32H67O4P/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-35-37(33,34)36-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-32H2,1-2H3,(H,33,34)

InChI key

RNPXCFINMKSQPQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Dihexadecyl phosphate (DHP) is a negatively charged lipid that is used for the formation of model membranes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Susruta Samanta et al.
ACS infectious diseases, 4(10), 1519-1528 (2018-07-25)
Understanding molecular properties of outer membrane channels of Gram-negative bacteria is of fundamental significance as they are the entry point of polar antibiotics into bacteria. Outer membrane proteomics revealed OccK8 (OprE) to be among the five most expressed substrate specific
Y Ozeki et al.
Infection and immunity, 65(5), 1793-1799 (1997-05-01)
It is reported that some bacteria or bacterial components cause thymic atrophy via the apoptotic process. The present study demonstrated for the first time in vivo induction of apoptosis in the mouse thymus by mycobacterial cord factor (CF) (trehalose 6,6'-dimycolate).
Ba Hieu Tran et al.
International journal of nanomedicine, 14, 10061-10077 (2020-01-11)
S-propargyl-cysteine (SPRC; alternatively known as ZYZ-802) is a novel modulator of endogenous tissue H2S concentrations with known cardioprotective and anti-inflammatory effects. However, its rapid metabolism and excretion have limited its clinical application. To overcome these issues, we have developed some
Smita Pawar et al.
Journal of drug targeting, 24(8), 730-743 (2016-02-16)
Efficacy of anticancer drug is limited due to non-selectivity and toxicities allied with the drug; therefore the heart of the present work is to formulate drug delivery systems targeted selectively towards cancer cells with minimal toxicity to normal cells. Targeted
Haruyuki Ishii et al.
Colloids and surfaces. B, Biointerfaces, 92, 372-376 (2011-12-16)
Hollow silica particle was obtained with a vesicle template synthesis in water under ambient conditions in the presence of ammonia. Biomimetic vesicles, liposomes were used, which consisted of a zwitterionic phospholipid, 1,2-dipalmitoyl-sn-glycero-3-phosphocholine (DPPC), and a tiny amount of charged amphiphiles

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service