Skip to Content
Merck
All Photos(2)

Documents

D1916

Sigma-Aldrich

5,6-Dichlorobenzimidazole 1-β-D-ribofuranoside

≥98% (HPLC), powder, RNA synthesis inhibitor

Synonym(s):

5,6-Dichloro-1-β-D-ribofuranosylbenzimidazole, 5,6-Dichlorobenzimidazole riboside, DRB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12Cl2N2O4
CAS Number:
Molecular Weight:
319.14
Beilstein:
39123
MDL number:
UNSPSC Code:
12352200
eCl@ss:
32151902
PubChem Substance ID:
NACRES:
NA.77

product name

5,6-Dichlorobenzimidazole 1-β-D-ribofuranoside,

form

powder

Quality Level

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)n2cnc3cc(Cl)c(Cl)cc23

InChI

1S/C12H12Cl2N2O4/c13-5-1-7-8(2-6(5)14)16(4-15-7)12-11(19)10(18)9(3-17)20-12/h1-2,4,9-12,17-19H,3H2/t9-,10-,11-,12-/m1/s1

InChI key

XHSQDZXAVJRBMX-DDHJBXDOSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

5,6-Dichlorobenzimidazole 1-β-D-ribofuranoside has been used:
  • as an inhibitor of RNA polymerase II in mouse melanoma cells
  • for the inhibition of cyclin D1 mRNA synthesis in human prostate epithelial cell lines
  • in the inhibition of interleukin-2 gene transcription in Jurkat cells

Biochem/physiol Actions

5,6-Dichlorobenzimidazole 1-β-D-ribofuranoside (DRB), a nucleoside analog that halts mRNA synthesis by phosphorylation of the C-terminal domain of RNA polymerase II, making it inactive. It also interferes with the DNA topoisomerase II, may modulate response to cytokines and blocks the human immunodeficiency virus (HIV) via RNA modification. It also inhibits cyclin-dependent kinases (CDKs) 7 and 9 and favors apoptosis in leukemic cells. It may serve as a therapeutic agent in treating cancer.
Inhibitor of RNA synthesis; causes premature termination of transcription. CK2 (casein kinase-2) inhibitor.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Transcriptional regulation of interleukin-2 gene expression is impaired by copper deficiency in Jurkat human T lymphocytes
Hopkins RG and Failla ML
The Journal of Nutrition, 129(3), 596-601 (1999)
NELF, a multisubunit complex containing RD, cooperates with DSIF to repress RNA polymerase II elongation
Yamaguchi Y, et al.
Cell, 97(1), 41-51 (1999)
M Ljungman et al.
Oncogene, 18(3), 583-592 (1999-02-16)
The mechanisms by which the p53 response is triggered following exposure to DNA-damaging agents have not yet been clearly elucidated. We and others have previously suggested that blockage of RNA polymerase II may be the trigger for induction of the
Lilija Brant et al.
Molecular systems biology, 12(12), 891-891 (2016-12-13)
Mammalian interphase chromosomes fold into a multitude of loops to fit the confines of cell nuclei, and looping is tightly linked to regulated function. Chromosome conformation capture (3C) technology has significantly advanced our understanding of this structure-to-function relationship. However, all
Androgen receptor-mediated growth suppression of HPr-1AR and PC3-Lenti-AR prostate epithelial cells
Kim YC, et al.
PLoS ONE, 10(9), e0138286-e0138286 (2015)

Articles

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service