D1285
2′,3′-Dideoxyadenosine
≥97% (HPLC)
Synonym(s):
Dideoxyadenosine, ddA
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H13N5O2
CAS Number:
Molecular Weight:
235.24
Beilstein:
619924
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51
Recommended Products
Assay
≥97% (HPLC)
form
powder
optical activity
[α]/D -28.0±1.0
mp
181-184 °C (lit.)
solubility
water: 100 mg/mL, clear, colorless
storage temp.
2-8°C
SMILES string
Nc1ncnc2n(cnc12)[C@H]3CC[C@@H](CO)O3
InChI
1S/C10H13N5O2/c11-9-8-10(13-4-12-9)15(5-14-8)7-2-1-6(3-16)17-7/h4-7,16H,1-3H2,(H2,11,12,13)/t6-,7+/m0/s1
InChI key
WVXRAFOPTSTNLL-NKWVEPMBSA-N
Gene Information
human ... DCK(1633)
Looking for similar products? Visit Product Comparison Guide
General description
2′,3′-Dideoxyadenosine is a nucleoside analog of deoxyadenosine. On deamination, it is converted to dideoxyinosine. It inhibits adenylyl cyclase and may play a key role in the inhibition of tumor progression. It is a potent inhibitor of reverse transcriptase enzyme of human immunodeficiency virus.
Application
2′,3′-Dideoxyadenosine (ddA), a specific adenylyl cyclase inhibitor, is useful in biological process and pathway studies involving adenylyl cyclase activity and cAMP pool modulation.
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
James N Wilson et al.
Chembiochem : a European journal of chemical biology, 9(2), 279-285 (2007-12-12)
Fluorescent nucleosides are widely used as probes of biomolecular structure and mechanism in the context of DNA, but they often exhibit low quantum yields because of quenching by neighboring DNA bases. Here we characterize the quenching by DNA of fluorescent
Elena Grossini et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 23(1-3), 87-96 (2009-03-04)
Urocortin II has previously been shown in anesthetized pigs to increase coronary blood flow through activation of the endothelial nitric oxide synthase (eNOS) pathway and involvement of the subtype 2 of corticotropin releasing factor receptors (CRFR2). However, little information has
Christopher Tubbs et al.
Endocrinology, 150(1), 473-484 (2008-09-20)
Progestin stimulation of sperm hypermotility remains poorly understood despite having been described in numerous vertebrate species. We show here that progestin stimulation of sperm hypermotility in a teleost, the Atlantic croaker (Micropogonias undulatus) is associated with activation of an olfactory
Pharmacokinetics of 2′, 3′-dideoxyadenosine and 2′, 3′-dideoxyinosine in patients with severe human immunodeficiency virus infection
Hartman NR, et al.
Clinical Pharmacology and Therapeutics, 47(5), 647-654 (1990)
Mark Kwaaitaal et al.
The Biochemical journal, 440(3), 355-365 (2011-08-19)
Binding of specific microbial epitopes [MAMPs (microbe-associated molecular patterns)] to PRRs (pattern recognition receptors) and subsequent receptor kinase activation are key steps in plant innate immunity. One of the earliest detectable events after MAMP perception is a rapid and transient
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service