Skip to Content
Merck
All Photos(1)

Key Documents

284386

Sigma-Aldrich

N,N-Dimethyldodecylamine

97%

Synonym(s):

1-(Dimethylamino)dodecane, DDA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11N(CH3)2
CAS Number:
Molecular Weight:
213.40
Beilstein:
1700191
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

<129 mmHg ( 21 °C)

Assay

97%

form

liquid

refractive index

n20/D 1.4375 (lit.)

bp

80-82 °C/0.1 mmHg (lit.)

mp

−20 °C (lit.)

density

0.787 g/mL at 20 °C (lit.)

functional group

amine

SMILES string

CCCCCCCCCCCCN(C)C

InChI

1S/C14H31N/c1-4-5-6-7-8-9-10-11-12-13-14-15(2)3/h4-14H2,1-3H3

InChI key

YWFWDNVOPHGWMX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

N,N-Dimethyldodecylamine has been used:
  • as a capping agent during the synthesis of Au(core)-Pd(shell) bimetallic nanoparticles in toluene
  • in the synthesis of N-alkyl-N,N-dimethyl-1-ammonio-2-hydroxy-3-propane sulfonate
  • in the preparation of novel bis-quaternary ammonium salt, via reaction with epichlorohydrin (in the presence of the amine hydrochloride) or various dichloro compounds

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sudip Nath et al.
Langmuir : the ACS journal of surfaces and colloids, 21(23), 10405-10408 (2005-11-03)
In this paper a convenient route for synthesizing Au(core)-Pd(shell) bimetallic nanoparticles in toluene has been reported as a result of co-reduction of gold(III) and palladium(II) precursors in toluene. N,N-Dimethyldodecylamine was used as a capping agent for the core-shell particles, which
Marc van Oostrum et al.
Nature communications, 11(1), 4990-4990 (2020-10-07)
Neurons are highly compartmentalized cells with tightly controlled subcellular protein organization. While brain transcriptome, connectome and global proteome maps are being generated, system-wide analysis of temporal protein dynamics at the subcellular level are currently lacking. Here, we perform a temporally-resolved
Nanika Coetzee et al.
Scientific reports, 7(1), 607-607 (2017-04-06)
Gene expression in Plasmodia integrates post-transcriptional regulation with epigenetic marking of active genomic regions through histone post-translational modifications (PTMs). To generate insights into the importance of histone PTMs to the entire asexual and sexual developmental cycles of the parasite, we
Britta Eggenreich et al.
BMC biotechnology, 18(1), 30-30 (2018-05-31)
Celiac disease (CD) is one of the most common food-related chronic disorders. It is mediated by the dietary consumption of prolamins, which are storage proteins of different grains. So far, no therapy exists and patients are bound to maintain a
Sanjay Shahi et al.
Biology, 9(5) (2020-05-10)
Currently, 5-fluorouracil (5-FU)-based combination chemotherapy is the mainstay in the treatment of metastatic colorectal cancer (CRC), which benefits approximately 50% of the patients. However, these tumors inevitably acquire chemoresistance resulting in treatment failure. The molecular mechanisms driving acquired chemotherapeutic drug

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service