Skip to Content
Merck
All Photos(1)

Key Documents

D1260

Sigma-Aldrich

Decamethonium bromide

crystalline

Synonym(s):

Decamethylene bis(trimethylammonium bromide), Decane-1,10-bis(trimethylammonium bromide)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3N(Br)(CH2)10N(Br)(CH3)3
CAS Number:
Molecular Weight:
418.29
Beilstein:
3728288
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

form

crystalline

Quality Level

color

off-white

mp

263-267 °C (lit.)

originator

GlaxoSmithKline

SMILES string

[Br-].[Br-].C[N+](C)(C)CCCCCCCCCC[N+](C)(C)C

InChI

1S/C16H38N2.2BrH/c1-17(2,3)15-13-11-9-7-8-10-12-14-16-18(4,5)6;;/h7-16H2,1-6H3;2*1H/q+2;;/p-2

InChI key

HLXQFVXURMXRPU-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Decamethonium bromide has been used to induce paralysis in duck embryo.[1]

Biochem/physiol Actions

Decamethonium serves as a muscle relaxant and is also a neuromuscular blocking agent. It has a molecular weight of 258.4.[2]
Nicotinic acetylcholine receptor partial agonist and neuromuscular blocking agent; depolarizes striated muscles and blocks their activity.

Features and Benefits

This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Mesenchymal and mechanical mechanisms of secondary cartilage induction
Solem R C, et al.
Developmental Biology, 356(1), 28-39 (2011)
D Bertrand et al.
Proceedings of the National Academy of Sciences of the United States of America, 87(5), 1993-1997 (1990-03-01)
Neuronal nicotinic acetylcholine receptor of the alpha 4/non-alpha (alpha 4/n alpha) type was reconstituted in Xenopus oocytes after nuclear injection of cDNA expression vectors. Functional neuronal receptor was only formed when the two subunits alpha 4 and n alpha were
Dana B Finley et al.
Journal of pharmacological and toxicological methods, 59(2), 108-119 (2009-04-16)
Exposure to irreversible cholinesterase (ChE)-inhibiting compounds, such as organophosphates may produce neuromuscular dysfunction. However, less is known about changes in neuromuscular transmission after treatment with reversible ChE-inhibitors. These studies adapted single fiber electromyography (SFEMG) techniques to quantify neuromuscular jitter in
I Wenningmann et al.
Molecular pharmacology, 60(3), 584-594 (2001-08-15)
We performed macroscopic and single-channel current measurements on wild-type (WT) and two mutant muscle-type nicotinic acetylcholine (ACh) receptor channels transiently expressed in HEK-293 cells. The mutants contained polar-to-nonpolar substitutions at the 10' (alpha(2)S10'A beta T10'A gamma delta) and 6' positions
Chigusa Sawamura et al.
Magnetic resonance in medicine, 56(6), 1235-1241 (2006-11-08)
To examine the effect of immobilization on the development of articular cartilage, we assessed glycosaminoglycan (GAG) content in the chick articular surface by delayed gadolinium-enhanced MRI of cartilage (dGEMRIC). Chick embryos were paralyzed by decamethonium bromide (DMB) from day 10

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service