CLS3875
Corning® 96 Well Half-Area Microplate
flat bottom, black polystyrene, Tissue Culture (TC)-treated surface, bag of 20, sterile, lid
Synonym(s):
cell culture plate, multi well plates, multiple well plates
About This Item
black polystyrene
flat bottom
round wells
Recommended Products
material
black bottom
black polystyrene
flat bottom
round wells
sterility
sterile
feature
lid
skirt
packaging
bag of 20
manufacturer/tradename
Corning 3875
size
96 wells
well D
10.7 mm
well volume
190 μL
well working volume
25-125 μL
color
black
suitability
suitable for tissue
binding type
Tissue Culture (TC)-treated surface
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Features and Benefits
- Corning® 96 Well Half-Area Microplate Flat bottoms with opaque walls enable minimal well-to-well crosstalk.
- 190 μL total volume and nonpyrogenic
- Reduced contamination aided by nonreversible lids with condensation rings
- Sterilized by gamma radiation
- Enables minimal background fluorescence and/or luminescence
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service