Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
96 Well Half-Area Microplate
Products
Product Category
plates (22)
Color
Manufacturer
Binding Type
Diameter (mm)
Volume (µL)
Wells
Color
Package Size
Sterility
Filter Products
Showing 1-20 of 22
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
CLS3690
Corning
®
96 Well Half-Area Microplate,
flat bottom clear, polystyrene, High, bag of 25 inner package, non-sterile, lid: no
Expand
View Pricing
CLS3694
Corning
®
96 Well Half-Area Microplate,
flat bottom, black polystyrene, bag of 25, non-sterile, lid: no
Expand
View Pricing
CLS3882
Corning
®
96 Well Half-Area Microplate,
flat bottom clear, black polystyrene, Tissue Culture (TC)-treated surface, bag of 20, sterile, lid
Expand
View Pricing
CLS3693
Corning
®
96 Well Half-Area Microplate,
flat bottom, white polystyrene, bag of 25 inner package, non-sterile, lid: no
Expand
View Pricing
CLS3686
Corning
®
96 Well Half-Area Microplate,
flat bottom, black polystyrene, NBS
™
(non-binding surface), bag of 25 inner package, non-sterile, lid: no
Expand
View Pricing
CLS3695
Corning
®
96 Well Half-Area Microplate,
flat bottom clear, polystyrene, bag of 25, non-sterile, lid: no
Expand
View Pricing
CLS3881
Corning
®
96 Well Half-Area Microplate,
flat bottom clear, black polystyrene, NBS
™
(non-binding surface), bag of 25, non-sterile, lid: no
Expand
View Pricing
CLS3992
Corning
®
96 Well Half-Area Microplate,
flat bottom, white polystyrene, NBS
™
(non-binding surface), bag of 5, non-sterile, lid: no
Expand
View Pricing
CLS3993
Corning
®
96 Well Half-Area Microplate,
flat bottom, black polystyrene, NBS
™
(non-binding surface), bag of 5, non-sterile, lid: no
Expand
View Pricing
CLS3642
Corning
®
96 Well Half-Area Microplate,
flat bottom, white polystyrene, bag of 25 inner package, non-sterile, lid: no
Expand
View Pricing
CLS4680
Corning
®
96 Well Half-Area Microplate,
High Content Imaging Film bottom, sterile, lid
Expand
View Pricing
CLS3885
Corning
®
96 Well Half-Area Microplate,
flat bottom clear, white polystyrene, Tissue Culture (TC)-treated surface, bag of 20, sterile, lid
Expand
View Pricing
CLS3696
Corning
®
96 Well Half-Area Microplate,
flat bottom clear, Tissue Culture (TC)-treated surface, pkg of (individually wrapped), pkg of (50/case), sterile, lid
Expand
View Pricing
CLS3880
Corning
®
96 Well Half-Area Microplate,
flat bottom clear, black polystyrene, bag of 25, non-sterile, lid: no
Expand
View Pricing
CLS3688
Corning
®
96 Well Half-Area Microplate,
flat bottom, white polystyrene, Tissue Culture (TC)-treated surface, bag of 20 inner package, sterile, lid
Expand
View Pricing
CLS3875
Corning
®
96 Well Half-Area Microplate,
flat bottom, black polystyrene, Tissue Culture (TC)-treated surface, bag of 20, sterile, lid
Expand
View Pricing
CLS3697
Corning
®
96 Well Half-Area Microplate,
flat bottom clear, Tissue Culture (TC)-treated surface, bag of 20, sterile, lid
Expand
View Pricing
CLS3886
Corning
®
96 Well Half-Area Microplate,
flat bottom clear, white polystyrene, Tissue Culture (TC)-treated surface, bag of 20, sterile, lid: no
Expand
View Pricing
CLS3994
Corning
®
96 Well Half-Area Microplate,
flat bottom clear, white polystyrene, NBS
™
(non-binding surface), bag of 5, non-sterile, lid: no
Expand
View Pricing
CLS3887
Corning
®
96 Well Half-Area Microplate,
flat bottom clear, black polystyrene, Tissue Culture (TC)-treated surface, bag of 25, sterile, lid: no
Expand
View Pricing
Page 1 of 2
Page 1 of 2