Skip to Content
Merck
All Photos(1)

Key Documents

CG201

Sigma-Aldrich

cGMP Enzyme Immunoassay Kit

sufficient for 96 assays

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.32

usage

sufficient for 96 assays

Quality Level

technique(s)

ELISA: suitable

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... PRKG1(5592)

General description

cGMP Enzyme Immunoassay Kit is non-radioactive, competitive immunoassay for the quantitation of cyclic guanine monophosphate (cGMP) in biological fluids (serum, plasma, urine and saliva), tissue culture media samples or in samples containing very low concentrations of cyclic nucleotides. This kit uses a polyclonal antibody to cGMP to competitively bind cGMP or cGMP that has been covalently linked to an alkaline phosphatase molecule. The immunoassay is performed in a 96 well plate coated with anti-rabbit IgG antibody. The colored end product, produced by the addition of substrate to the wells, is read at 405 nm on a multiwell plate reader. The intensity of the color is inversely proportional to the concentration of cGMP present in the well.

Application

Non-radioactive, competitive immunoassay for the quantitation of cGMP in biological fluids (serum, plasma, urine and saliva), tissue culture media samples, or in samples containing very low concentrations of cyclic nucleotides. This kit uses a polyclonal antibody to cGMP to competitively bind cGMP or cGMP that has been covalently linked to an alkaline phosphatase molecule. The assay is performed in a 96 well plate coated with anti-rabbit IgG antibody. The colored end product, produced by the additon of substrate to the wells, is read at 405 nm on a multiwell plate reader. The intensity of the color is inversely proportional to the concentration of cGMP present in the well.
cGMP Enzyme Immunoassay Kit has been used for the quantitative determination of cyclic guanine monophosphate (cGMP) concentration in parasite tissues and mouse aortic artery homogenates. It has also been used to assess the guanylate cyclase activity.

Kit Components Only

Product No.
Description

  • Acetic Anhydride 1 x 1

  • Assay Buffer 2 1 x 30

  • cGMP-Alkaline Phosphatase Conjugate 1 x 5

  • cGMP Assay Layout Sheet 1 x 1

  • cGMP EIA Antibody Rabbit Anti-cGMP 1 x 5

  • 5 Cycle Logit-Log Paper 1 x 1

  • Cyclic GMP Standard 1 x 0.5

  • Goat Anti-Rabbit IgG Coated 96 Well Microtiter Plate 1 ea

  • p-Nitrophenyl Phosphate Substrate Solution 1 x 20

  • Plate Sealer 1 ea

  • Stop Solution 1 x 5

  • Triethylamine 1 x 2

  • Wash Buffer Concentrate 1 x 30

See All (13)

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

Flash Point(F)

12.2 °F - closed cup

Flash Point(C)

-11 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Identification of a novel Arabidopsis thaliana nitric oxide-binding molecule with guanylate cyclase activity in vitro
Mulaudzi T, et al.
Febs Letters, 585(17), 2693-2697 (2011)
Inhibition of MPO (Myeloperoxidase) Attenuates Endothelial Dysfunction in Mouse Models of Vascular Inflammation and Atherosclerosis
Cheng D, et al.
Arteriosclerosis, Thrombosis, and Vascular Biology, 39(7), 1448-1457 (2019)
Genistein from Flemingia vestita (Fabaceae) enhances NO and its mediator (cGMP) production in a cestode parasite, Raillietina echinobothrida
Das B, et al.
Parasitoloty, 134(10), 1457-1463 (2007)
Sara Falvo et al.
Animals : an open access journal from MDPI, 11(6) (2021-07-03)
The quail Coturnix coturnix is a seasonal breeding species, with the annual reproductive cycle of its testes comprising an activation phase and a regression phase. Our previous results have proven that the testicular levels of both 17β-estradiol (E2) and androgens
Kamal Kumar Malukani et al.
Plant physiology, 183(3), 1345-1363 (2020-05-02)
Plant pathogens secrete cell wall-degrading enzymes that degrade various components of the plant cell wall. Plants sense this cell wall damage as a mark of infection and induce immune responses. However, the plant functions that are involved in the elaboration

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service