Skip to Content
Merck
All Photos(1)

Key Documents

AB1 cell line from mouse lung

10092305, mouse lung. Fibroblast-like, but demonstrates typical epithelial cobblestone morphology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

AB1 cell line from mouse lung, 10092305, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 10092305, mouse lung. Fibroblast-like, but demonstrates typical epithelial cobblestone morphology

biological source

mouse lung

description

Mouse malignant mesothelioma

packaging

tube of 5 μg 10092305-DNA-5UG
pkg of vial of cells 10092305-1VL

growth mode

Adherent

karyotype

Not specified

morphology

Fibroblast-like but shows a typical epithelial "cobblestone"

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse malignant mesothelioma

Cell Line Description

Mice (BALB/c female 6-8 weeks) were exposed to crocidolite asbestos through intraperitoneal injection resulting in tumour development. Cultures were established from malignant mesothelial cells obtained from ascites fluid. Cells are tumourigenic in syngeneic immunocompetent mice. Cell Bank Australia reference number: CBA0144

Application

Investigation of the properties of malignant mesothelioma cells and the development of new diagnostic methods.

Culture Medium

RPMI 1640 (with 2mM L-Glutamine + 25mM HEPES) + 5% FCS

Subculture Routine

Optimal split ratio 1:8 (seeding density 1.2 x104 cells/cm2).Cells incubated at 37oC with 5% CO2.Harvest the cells using 0.05% Trypsin/EDTA at 37°C for 5 min.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments.  Please check with your local Customer Service representative for more information.
Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.
Depositor and originator: Richard Lake, University of Western Australia, Australia

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service