Skip to Content
Merck
All Photos(2)

Key Documents

SCC145

Sigma-Aldrich

ID8 Mouse Ovarian Surface Epithelial Cell Line

Mouse

Synonym(s):

ID-8, ID8/MOSEC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.81
Pricing and availability is not currently available.

Product Name

ID8 Mouse Ovarian Surface Epithelial Cell Line, ID8 mouse ovarian surface epithelial cell line is frequently used as a syngeneic mouse model for human ovarian cancer.

biological source

mouse

technique(s)

cell culture | mammalian: suitable

shipped in

liquid nitrogen

General description

ID8 is one of 10 clonal lines established from late passaged C57BL/6 murine ovarian surface epithelial cells (MOSEC) [1]. Intraperitoneal injection of each of the 10 clonal lines into C57BL/6 mice resulted in formation of peritoneal tumors and ascitic fluid. Of the 10 clonal lines, ID8 exhibited the highest tumor load. ID8 cell line is a highly published and well characterized cell line and is frequently used as a syngeneic mouse model for ovarian cancer.
Ovarian cancer is the fourth leading cause of cancer-related deaths in women. The disease is frequently diagnosed at later stages, with tumors detected throughout the peritoneal cavity. Approximately 90% of ovarian tumors arise from ovarian surface epithelial cells [2]. Human and mouse ovarian surface epithelial cells (OSE) have been isolated and are used to develop ovarian cancer models. These models typically involve injection of the human/mouse OSE cells subcutaneously, intraperitoneally or orthotopically into immune deficient mice. A common drawback to these models is the absence of an intact immune system in the host mice.

In 2000, an immune-competent syngeneic mouse model for ovarian cancer was reported [1]. Mouse ovarian surface epithelial cells (MOSEC) isolated from C57BL/6 mice were found to spontaneously transform into malignant tumorigenic cells following prolonged passages in vitro [1]. Late passage MOSEC lost the classical “cobblestone” contact-inhibited in vitro properties reminiscent of normal epithelial cells and instead grew as multi-layered cell clusters indicative of transformed cells. Intraperitoneal injection of late passaged MOSEC into athymic and normal, immune-intact, syngeneic C57BL/6 mice gave rise to tumors throughout the abdominal cavity like those observed in women with Stage III and IV cancer [1]. MOSEC are thus useful syngeneic mouse models to study the role of the immune system in the establishment and progression of ovarian cancer.

Quality

• Each vial contains ≥ 1X106 viable cells.
• Cells are tested negative for infectious diseases by a Mouse Essential CLEAR panel by Charles River Animal Diagnostic Services.
• Cells are verified to be of mouse origin and negative for inter-species contamination from rat, chinese hamster, Golden Syrian hamster, human and non-human primate (NHP) as assessed by a Contamination CLEAR panel by Charles River Animal Diagnostic Services.
• Cells are negative for mycoplasma contamination

Other Notes

This product is intended for sale and sold solely to academic institutions for internal academic research use per the terms of the "Academic Use Agreement" as detailed in the product documentation. For information regarding any other use, please contact [email protected].

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

James Greenaway et al.
Gynecologic oncology, 108(2), 385-394 (2007-11-27)
Ovarian cancer is the fourth leading cause of cancer-related deaths among women and is among the least understood of all cancers. The objective of this study was to determine the effect of ovarian epithelial and stromal cell interaction in a
Amr Khalifa et al.
Nature communications, 14(1), 6951-6951 (2023-11-01)
Identifying oncological applications for drugs that are already approved for other medical indications is considered a possible solution for the increasing costs of cancer treatment. Under the hypothesis that nutritional stress through fasting might enhance the antitumour properties of at
Qian-Feng Zhang et al.
Theranostics, 10(23), 10619-10633 (2020-09-16)
Great progress has been made in the field of tumor immunotherapy in the past decade. However, the therapeutic effects of immune checkpoint blockade (ICB) against ovarian cancer are still limited. Recently, an inhibitor of cyclin-dependent kinases 4 and 6 (CDK4/6i)
Wangyou Feng et al.
Heliyon, 9(9), e19760-e19760 (2023-10-09)
Ovarian cancer is insensitive to immunotherapy and has a high mortality rate. CDK4/6 inhibitors (CDK4/6i) regulate the tumor microenvironment and play an antitumor role. Our previous research demonstrated that lymphocyte aggregation (tertiary lymphoid structures, TLSs) was observed after CDK4/6i treatment.
K F Roby et al.
Carcinogenesis, 21(4), 585-591 (2000-04-07)
Mouse ovarian surface epithelial cells (MOSEC) were obtained from virgin, mature mice by mild trypsinization and were repeatedly passaged in vitro. Early passage cells (<20 passages) exhibited a cobblestone morphology and contact inhibition of growth. After approximately 20 passages in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service