Skip to Content
Merck
All Photos(1)

Documents

C9705

Sigma-Aldrich

Concanamycin A

≥70% (HPLC), crystals, vacuolar-type v-ATPase inhibitor

Synonym(s):

Folimycin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H75NO14
CAS Number:
Molecular Weight:
866.09
Beilstein:
3560277
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

Concanamycin A, ≥70% (HPLC)

Quality Level

Assay

≥70% (HPLC)

form

solid film

antibiotic activity spectrum

viruses

Mode of action

enzyme | inhibits

storage temp.

−20°C

SMILES string

CCC1C(O)C(C)C\C(C)=C\C=C\C(OC)C(OC(=O)\C(OC)=C\C(C)=C\C(C)C1O)C(C)C(O)C(C)[C@]2(O)C[C@@H](O[C@H]3C[C@@H](O)[C@H](OC(N)=O)[C@@H](C)O3)[C@H](C)C(O2)\C=C\C

InChI

1S/C46H75NO14/c1-13-16-34-28(7)37(58-38-22-33(48)43(31(10)57-38)60-45(47)53)23-46(54,61-34)30(9)41(51)29(8)42-35(55-11)18-15-17-24(3)19-26(5)39(49)32(14-2)40(50)27(6)20-25(4)21-36(56-12)44(52)59-42/h13,15-18,20-21,26-35,37-43,48-51,54H,14,19,22-23H2,1-12H3,(H2,47,53)/b16-13+,18-15+,24-17+,25-20+,36-21-/t26?,27?,28-,29?,30?,31-,32?,33-,34?,35?,37-,38+,39?,40?,41?,42?,43-,46+/m1/s1

InChI key

DJZCTUVALDDONK-LWLXYWDNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: macrolide
Concanamycin A belongs to the plecomacrolide family and comprises an 18-membered tetraenic macrolide ring.

Application

Concanamycin A has been used:
  • as a lysosomal inhibitor in young and old fibroblasts
  • as a vacuolar-type H+-ATPase inhibitor in presynaptic vesicles
  • as a lysosomal acidification blocker in HepG2 hepatocytes cells

Biochem/physiol Actions

Concanamycin A (ConA) inhibits acidification of organelles and perforin-mediated cytotoxicity. It is a vacuolar-type v-ATPase inhibitor. ConA possesses antiprotozoal and antineoplastic properties. It mediates inhibition of the negative factor (Nef) protein of the human immunodeficiency virus.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral - Eye Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christiane Ott et al.
Redox biology, 10, 266-273 (2016-11-09)
The overall decrease in proteolytic activity in aging can promote and accelerate protein accumulation and metabolic disturbances. To specifically analyze changes in macroautophagy (MA) we quantified different autophagy-related proteins (ATGs) in young, adult and old murine tissue as well as
Stephen F Haydock et al.
Microbiology (Reading, England), 151(Pt 10), 3161-3169 (2005-10-07)
The macrolide antibiotic concanamycin A has been identified as an exceptionally potent inhibitor of the vacuolar (V-type) ATPase. Such compounds have been mooted as the basis of a potential drug treatment for osteoporosis, since the V-ATPase is involved in the
Hideaki Toda et al.
Developmental and comparative immunology, 35(1), 88-93 (2010-09-04)
T cell-mediated cytotoxicity occurs via pathways based on perforin or Fas mechanisms. Perforin is a protein present in the cytoplasmic granules of CD8(+) cytotoxic T lymphocytes and is secreted to form pores on target cell membranes. In fish, although the
Mark M Painter et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(38), 23835-23846 (2020-09-10)
Nef is an HIV-encoded accessory protein that enhances pathogenicity by down-regulating major histocompatibility class I (MHC-I) expression to evade killing by cytotoxic T lymphocytes (CTLs). A potent Nef inhibitor that restores MHC-I is needed to promote immune-mediated clearance of HIV-infected
Jörn Dengjel et al.
Molecular & cellular proteomics : MCP, 11(3), M111-M111 (2012-02-09)
Autophagy is one of the major intracellular catabolic pathways, but little is known about the composition of autophagosomes. To study the associated proteins, we isolated autophagosomes from human breast cancer cells using two different biochemical methods and three stimulus types:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service