Skip to Content
Merck
All Photos(1)

Key Documents

D7693

Sigma-Aldrich

Dynasore hydrate

≥97.5% (HPLC), powder, dynamin 1 and dynamin 2 GTPase activity inhibitor

Synonym(s):

3-Hydroxy-naphthalene-2-carboxylic acid (3,4-dihydroxy-benzylidene)-hydrazide hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H14N2O4 · xH2O
CAS Number:
Molecular Weight:
322.31 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

Dynasore hydrate,

form

powder

Quality Level

storage condition

protect from light

color

off-white

solubility

DMSO: ≥10 mg/mL
H2O: insoluble

storage temp.

−20°C

SMILES string

O.Oc1ccc(\C=N\NC(=O)c2cc3ccccc3cc2O)cc1O

InChI

1S/C18H14N2O4.H2O/c21-15-6-5-11(7-17(15)23)10-19-20-18(24)14-8-12-3-1-2-4-13(12)9-16(14)22;/h1-10,21-23H,(H,20,24);1H2/b19-10+;

InChI key

FKWZHQSDBMSHTN-ZIOFAICLSA-N

General description

Dynasore is a small molecule GTPase inhibitor that targets dynamin-1, dynamin-2 and Drp1 (mitochondrial). Dynasore blocks dynamin-dependent endocytosis, scission of endocytic vesicles. Dynosore is profibrotic, induces PAI-1 in pleural mesothelial cells, and may be an effective pleurodesing (pleurodesis) agent. Dynasor blocks human papillomavirus type 16 (HPV16) and bovine papillomavirus type 1 (BPV1) (pseudovirions) infections.
Dynasore hydrate lowers labile cholesterol in the plasma membrane and distorts lipid raft organization. Dynasore exhibits pleotropic effects on herpes simplex virus (HSV)-1 and HSV-2 infection. It hinders viral entry, trafficking of viral proteins and capsid formation.

Application

Dynasore hydrate has been used:
  • for priming fetal mesenchymal stem/stromal cells (fMSC)
  • in confocal microscopy
  • in wound healing/scratch and tube-formation assay
  • to analyse its effects on Coxsackievirus A9 (CAV9) infection
  • to inhibit clathrin-mediated endocytosis and caveolin-mediated endocytosis
  • in live imaging of cells

Biochem/physiol Actions

Dynasore is a cell-permeable, reversible noncompetitive dynamin 1 and dynamin 2 GTPase activity inhibitor.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Intracellular trafficking and endocytosis of CXCR4 in fetal mesenchymal stem/stromal cells
Pelekanos RA, et al.
BMC Cell Biology, 15(1), 15-15 (2014)
Dynasore-not just a dynamin inhibitor
Preta G, et al.
Cell communication and signaling : CCS, 13(1), 24-24 (2015)
Florian Ehlgen et al.
PloS one, 7(6), e38781-e38781 (2012-06-22)
Haemoglobin degradation during the erythrocytic life stages is the major function of the food vacuole (FV) of Plasmodium falciparum and the target of several anti-malarial drugs that interfere with this metabolic pathway, killing the parasite. Two multi-spanning food vacuole membrane
Tom Kirchhausen et al.
Methods in enzymology, 438, 77-93 (2008-04-17)
The large GTPase dynamin is essential for clathrin-dependent coated-vesicle formation. Dynasore is a cell-permeable small molecule that inhibits the GTPase activity of dynamin1, dynamin2 and Drp1, the mitochondrial dynamin. Dynasore was discovered in a screen of approximately 16,000 compounds for
G J Hofmeyr et al.
BJOG : an international journal of obstetrics and gynaecology, 121(8), 951-957 (2014-03-14)
Epidemiological data link low dietary calcium with pre-eclampsia. Current recommendations are for 1.5-2 g/day calcium supplementation for low-intake pregnant women, based on randomised controlled trials of ≥1 g/day calcium supplementation from 20 weeks of gestation. This is problematic logistically in

Articles

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service