Skip to Content
Merck
All Photos(1)

Key Documents

C6854

Sigma-Aldrich

Cathepsin L from human liver

≥0.5 units/mg protein, solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32
Pricing and availability is not currently available.

biological source

human liver

Quality Level

form

solution

specific activity

≥0.5 units/mg protein

technique(s)

activity assay: suitable

pH range

4.5—5.5

UniProt accession no.

application(s)

pharmaceutical

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... CTSL1(1514)

General description

The cathepsin L (CTSL) gene is mapped to human chromosome 9q21.33. It encodes a lysosomal cysteine proteinase that belongs to the peptidase C1 family. The enzyme is a dimer of a heavy (175 amino acids) and a light chain (42 amino acids) linked by disulfide bonds.

Application

Cathepsin L (CTSL) from human liver has been used:
  • to digest mutant transforming growth factor β-induced (TGFBIp) and LC3 (autophagosomes marker) proteins in vitro
  • to digest the pH-dependent fibril of a pre-melanosomal protein (Pmel17) repeat domain (RPT) isoform and study the effect of pH on sRPT aggregation kinetics by tryptophan fluorescence
  • to analyze the inhibition of CTSL by small molecules or drugs in a cell-free system

Biochem/physiol Actions

Cathepsin L (CTSL) exhibits endopeptidase activity and breaks peptide bonds with aromatic residues. This enzyme is functional at pH 3.0–6.5 with thiol compounds. Elastin, collagen, and α-1 protease inhibitors act as substrates for cathepsin L (CTSL). CTSL cleaves several proteins including enzymes, receptors, and transcription factors. It is involved in antigenic peptide production and controls B-cells homeostasis. It is associated with myofibril necrosis and tumor progression. Increased expression of CTSL promotes metastasis and relates to poor prognosis in cancer patients. Increased expression of CTSL is observed in breast cancer. CTSL is implicated in glioblastoma multiforme (GBM), middle east respiratory syndrome (MERS), gingival overgrowth, and severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2). It has a higher specific activity than cathepsin B and H in the degradation of a variety of physiological protein substrates.

Unit Definition

One unit will hydrolyze 1.0 μmole of Z-Phe-Arg-AFC per minute at pH 5.5 at 25 °C.

Physical form

Solution in in 20 mM malonate, pH 5.5, 1 mM EDTA, and 400 mM NaCl.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bing-Jie Lv et al.
Atherosclerosis, 230(1), 100-105 (2013-08-21)
Cathepsin L (CatL), cathepsin K (CatK), and cathepsin V (CatV) are potent elastases implicated in human arterial wall remodeling. Whether plasma levels of these cathepsins are altered in patients with abdominal aortic aneurysms (AAAs) remains unknown. Plasma samples were collected
Seung-Il Choi et al.
Journal of cellular and molecular medicine, 24(18), 10343-10355 (2020-07-16)
Granular corneal dystrophy type 2 (GCD2) is the most common form of transforming growth factor β-induced (TGFBI) gene-linked corneal dystrophy and is pathologically characterized by the corneal deposition of mutant-TGFBIp. The defective autophagic degradation of pathogenic mutant-TGFBIp has been shown
Stefan Tholen et al.
Cellular and molecular life sciences : CMLS, 71(5), 899-916 (2013-07-03)
Endolysosomal cysteine cathepsins functionally cooperate. Cathepsin B (Ctsb) and L (Ctsl) double-knockout mice die 4 weeks after birth accompanied by (autophago-) lysosomal accumulations within neurons. Such accumulations are also observed in mouse embryonic fibroblasts (MEFs) deficient for Ctsb and Ctsl.
Violeta Morin et al.
PloS one, 7(11), e46850-e46850 (2012-11-13)
Proteolysis of sperm histones in the sea urchin male pronucleus is the consequence of the activation at fertilization of a maternal cysteine protease. We previously showed that this protein is required for male chromatin remodelling and for cell-cycle progression in
Bailey Miller et al.
Journal of natural products, 77(1), 92-99 (2013-12-25)
A number of marine natural products are potent inhibitors of proteases, an important drug target class in human diseases. Hence, marine cyanobacterial extracts were assessed for inhibitory activity to human cathepsin L. Herein, we have shown that gallinamide A potently

Questions

  1. Hello, What is the protocol to bring this up in solution to have a concentration of 0.5 units/µL? Does units mean µg where the amount needed for performing digestions? Is there a protocol associated with this?

    1 answer
    1. This product is supplied as a solution. The minimum activity for each lot will be ≥0.5 units/mg protein. The specific activity of each lot is reported on the Certificate of Analysis. See the link below to review a sample Certificate:
      https://www.sigmaaldrich.com/certificates/sapfs/PROD/sap/certificate_pdfs/COFA/Q14/C6854-25UGSLCR2105.pdf

      The product is supplied in 20 mM malonate buffer, pH 5.5, with 1 mM EDTA and 400 mM NaCl. One unit is defined as the amount of Cathepsin L that hydrolyzes 1 uM Z-PHE-Arg-AFC per minute at 22 - 25 °C, in 400 mM Sodium Acetate, pH 5.5, containing 4 mM EDTA and 8 mM DTT.

      Please access the product datasheet at the link below:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/130/380/c6854dat.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service