Skip to Content
Merck
All Photos(2)

Key Documents

C9281

Sigma-Aldrich

Cholesterol Esterase from Pseudomonas fluorescens

lyophilized powder, ≥10,000 units/g protein

Synonym(s):

CE, bile salt-stimulated lipase, cholesteryl ester hydrolase, pancreatic cholesterol esterase, Sterol-ester acylhydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.

biological source

Pseudomonas fluorescens

Quality Level

Assay

10-30% (TCA-Biuret)

form

lyophilized powder

specific activity

≥10,000 units/g protein

mol wt

~129 kDa

composition

Protein, ~20%

storage condition

under inert gas (argon)

technique(s)

cell based assay: suitable

color

tan to brown

pH

7-9

solubility

0.4 M potassium phosphate, pH 7.0: soluble 1.0 mg/mL

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Research area: Cell Signaling

Cholesterol Esterase (CE) is a glycoprotein that can be isolated from fungal species such as Candida cylindracea and Pseudomonas fluorescens. It is classified as a member of the lipase/esterase family and functions as a homo-dimeric protein. CE is produced in the pancreas and is released in an active form upon stimulation by Cholecystokinin (CCK).[1]

Application

Cholesterol esterase from Pseudomonas fluorescens has been used in:
  • in cholesterol esterase assay to quantify total cholesterol from human blood serum samples[2]
  • a study to investigate the nondenaturing protein electro transfer of the esterase activity of lipolytic preparations
  • an optimization study of components in enzymatic cholesterol reagents containing cholesterol oxidase
  • for the modification of human plasma low-density-lipoprotein (LDL) to induce endothelial cell (EC) dysfunction and monocyte (MC) adhesion in the branched tissue-engineered blood vessels (TEBVs)[3]
  • to hydrolyze native cholesterol ester (CE) during ​filipin staining for detection of CE within the retinal frozen sections[4]
This enzyme is widely used in the determination of serum cholesterol in diagnostic laboratories.[5]

Biochem/physiol Actions

Cholesterol esterase (CE) is a reversible enzyme that can hydrolyze or synthesize fatty acid esters of cholesterol and other sterols. This activity of CE plays a vital role in steroid biosynthesis or de novo steroidogenesis.[6] Hydrolysis of water insoluble long chain fatty acid esters requires bile salt activation. Hydrolysis of water soluble esters of short chain fatty acids and lysophospholipids does not require activation by bile salts. It also hydrolyzes tri-, di-, and mono-acylglycerols, phospholipids, lysophospholipids, and ceramides. The enzyme may have multiple functions in lipid and lipoprotein metabolism, as well as in atherosclerosis.[7].

Other Notes

Contains potassium phosphate and TRITON® X-100.

Unit Definition

One unit will hydrolyze 1.0 μmole of cholesteryl oleate to cholesterol and oleic acid per min at pH 7.0 at 37 °C in the presence of taurocholate.

Analysis Note

Protein determined by biuret.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fat Digestion and Absorption
Viswanathan S, et al
Encyclopedia of Human Nutrition, 23-30 (2004)
Jounghyun H Lee et al.
Advanced biology, 5(4), e2000428-e2000428 (2021-04-15)
Atherosclerosis begins with the accumulation of cholesterol-carrying lipoproteins on blood vessel walls and progresses to endothelial cell dysfunction, monocyte adhesion, and foam cell formation. Endothelialized tissue-engineered blood vessels (TEBVs) have previously been fabricated to recapitulate artery functionalities, including vasoconstriction, vasodilation
Carolina Espinosa Álvarez et al.
Molecules (Basel, Switzerland), 25(24) (2020-12-30)
Haematococcus pluvialis is the largest producer of natural astaxanthin in the world. Astaxanthin is a bioactive compound used in food, feed, nutraceutics, and cosmetics. In this study, astaxanthin extraction from H. pluvialis by supercritical fluid extraction was evaluated. The effects
A colorimetric CMOS-based platform for rapid total serum cholesterol quantification
Al-Rawhani MA, et al.
IEEE Sensors Journal, 17, 240-247 (2016)
Syed A H Zaidi et al.
Journal of neuroinflammation, 20(1), 14-14 (2023-01-24)
We have investigated the efficacy of a new strategy to limit pathological retinal neovascularization (RNV) during ischemic retinopathy by targeting the cholesterol metabolizing enzyme acyl-coenzyme A: cholesterol transferase 1 (ACAT1). Dyslipidemia and cholesterol accumulation have been strongly implicated in promoting

Articles

Cholesterol esterification enhances transport efficiency in lipoproteins for increased blood stream transport.

Cholesterol esterification enhances transport efficiency in lipoproteins for increased blood stream transport.

Cholesterol esterification enhances transport efficiency in lipoproteins for increased blood stream transport.

Cholesterol esterification enhances transport efficiency in lipoproteins for increased blood stream transport.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service