Skip to Content
Merck
All Photos(3)

Documents

C8271

Sigma-Aldrich

Cytidine-5′-monophospho-N-acetylneuraminic acid sodium salt

≥85% (HPLC)

Synonym(s):

CMP-NAN, CMP-NANA, CMP-Neu5Ac, CMP-Sialic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C20H30N4NaO16P
CAS Number:
Molecular Weight:
636.43
Beilstein:
4224251
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

synthetic (organic)

Assay

≥85% (HPLC)

form

powder

storage temp.

−20°C

SMILES string

[Na+].CC(=O)N[C@@H]1[C@@H](O)C[C@@](O[C@@H]1[C@H](O)[C@H](O)CO)(OP([O-])(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N3C=CC(N)=NC3=O)C(O)=O

InChI

1S/C20H31N4O16P.Na/c1-7(26)22-12-8(27)4-20(18(32)33,39-16(12)13(29)9(28)5-25)40-41(35,36)37-6-10-14(30)15(31)17(38-10)24-3-2-11(21)23-19(24)34;/h2-3,8-10,12-17,25,27-31H,4-6H2,1H3,(H,22,26)(H,32,33)(H,35,36)(H2,21,23,34);/q;+1/p-1/t8-,9+,10+,12+,13+,14+,15+,16?,17+,20+;/m0./s1

InChI key

VFRHSOGUONIUOR-CTFMUGKASA-M

Looking for similar products? Visit Product Comparison Guide

General description

Cytosine 5′-monophosphate N-acetylneuraminic acid (CMP-Neu5Ac) is a sugar-nucleotide produced by CMP-Neu5Ac synthetase from CTP and Neu5Ac.

Application

Cytidine-5′-monophospho-N-acetylneuraminic acid sodium salt has been used:
  • as a standard in high-performance anion-exchange chromatography with pulsed amperometric detection (HPAEC-PAD) for nucleotide sugar analysis in Joubert syndrome type 10 (JBTS10) patient cells and control skin fibroblasts,
  • As a substrate for the enzymatic sialylation of G2 glycoforms, resialylation assay,
  • in in-vitro sialyltransferase assay

Biochem/physiol Actions

Cytidine-5′-monophospho-N-acetylneuraminic acid (CMP-Sialic acid) is a substrate for sialyltransferases. It is used for the enzymatic sialylation of glycans.
Cytosine 5′-monophosphate N-acetylneuraminic acid (CMP-Neu5Ac) is used as a substrate for Golgi sialyltransferases to form sialic acid and its conjugates.

Preparation Note

Enzymatically prepared by the method of Schauer, R., et al., Hoppe Seyler′s Z. Physiol. Chem., 353, 883 (1972).
CMP-NAN is very acid-labile.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yanhong Li et al.
Applied microbiology and biotechnology, 94(4), 887-905 (2012-04-25)
Sialic acids are a family of negatively charged monosaccharides which are commonly presented as the terminal residues in glycans of the glycoconjugates on eukaryotic cell surface or as components of capsular polysaccharides or lipooligosaccharides of some pathogenic bacteria. Due to
Francesca Necchi et al.
PloS one, 12(2), e0172163-e0172163 (2017-02-14)
Serum Bactericidal Activity (SBA) assay is the method of choice to evaluate the complement-mediated functional activity of both infection- and vaccine-induced antibodies. To perform a typical SBA assay, serial dilutions of sera are incubated with target bacterial strains and complement.
Christian Lizak et al.
Scientific reports, 7(1), 5842-5842 (2017-07-21)
Polysialic acid (polySia) is a homopolymeric saccharide that is associated with some neuroinvasive pathogens and is found on selective cell types in their eukaryotic host. The presence of a polySia capsule on these bacterial pathogens helps with resistance to phagocytosis
Gillian Dekkers et al.
Frontiers in immunology, 8, 877-877 (2017-08-22)
Glycosylation of the immunoglobulin G (IgG)-Fc tail is required for binding to Fc-gamma receptors (FcγRs) and complement-component C1q. A variety of IgG1-glycoforms is detected in human sera. Several groups have found global or antigen-specific skewing of IgG glycosylation, for example
Reassembled biosynthetic pathway for a large-scale synthesis of CMP-Neu5Ac
Song J, et al.
Marine drugs, 1(1), 34-45 (2003)

Articles

Explore tools for glycosyltransferase synthesis and modification of glycans, such as glycosyltransferases and nucleotide sugar donors.

Explore tools for glycosyltransferase synthesis and modification of glycans, such as glycosyltransferases and nucleotide sugar donors.

LC-MS/MS method quantifies similar polar nucleotide activated sugars using Supel™ Carbon LC column for simultaneous analysis.

LC-MS/MS method quantifies similar polar nucleotide activated sugars using Supel™ Carbon LC column for simultaneous analysis.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service