Skip to Content
Merck
All Photos(1)

Key Documents

S2563

Sigma-Aldrich

Sphingolipid Ceramide N-Deacylase from Pseudomonas sp.

Synonym(s):

SCDase

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352211
NACRES:
NA.32

foreign activity

exoglycosidases: α- and β-galactosidase, α- and β-N-acetylgalactosaminidase, β-N-acetylglucosaminidase, α-mannosidase, α-fucosidase, and sialidase., essentially free
protease and sphingomyelinase, essentially free

Quality Level

shipped in

dry ice

storage temp.

−20°C

General description

Sphingolipid Ceramide N-Deacylase from Pseudomonas sp. is a hydrolytic enzyme.

Application

Sphingolipid Ceramide N-Deacylase from Pseudomonas sp. has been used in enzymatic conversion and derivatization of sulfatides. It has also been used to hydrolyze the fatty acid chain of mactosyl ceramide.

Biochem/physiol Actions

Hydrolyzes the N-acyl linkage between fatty acids and sphingosines in ceramides of various sphingolipids.
Sphingolipid Ceramide N-Deacylase from Pseudomonas sp., under certain conditions, can recyclate lyso-sphingolipids. It condenses fatty acids to sphingosine to generate ceramide. The enzyme mainly acts on neutral and acidic glycosphingolipids.

Unit Definition

One unit will hydrolyze 1 μmol of asialo GM1 per minute at pH 6.0 at 37 °C.

Physical form

Solution in 50 mM sodium acetate, pH 6.0, with 0.1% Lubrol PX.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Kuchar et al.
Rapid communications in mass spectrometry : RCM, 24(16), 2393-2399 (2010-07-17)
Sphingolipid ceramide N-deacylase (SCDase, EC 3.5.1.69) is a hydrolytic enzyme isolated from Pseudomonas sp. TK 4. In addition to its primary deacylation function, this enzyme is able to reacylate lyso-sphingolipids under specific conditions. We immobilised this enzyme on magnetic macroporous
Characterization of the reversible nature of the reaction catalyzed by sphingolipid ceramide N-deacylase. A novel form of reverse hydrolysis reaction.
Kita K
European Journal of Biochemistry, 268(3), 592-602 (2001)
Sulfatide Analysis by Mass Spectrometry for Screening of Metachromatic Leukodystrophy in Dried Blood and Urine Samples.
Spacil Z
Clinical Chemistry, 62(1), 279-286 (2016)
D Thusitha U Abeytunga et al.
Bioorganic chemistry, 36(2), 70-76 (2007-11-21)
Glycosphingolipids (GSLs) have been implicated as playing major roles in cellular interactions and control of cell proliferation in muticellular organisms. Moreover GSLs and other sphingolipids such as sphingomyelins, ceramides and sphingosines serve a variety of roles in signal transduction. Hence
A novel enzyme that cleaves the N-acyl linkage of ceramides in various glycosphingolipids as well as sphingomyelin to produce their lyso forms.
Ito M
The Journal of Biological Chemistry, 270(41), 24370-24374 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service