Skip to Content
Merck
All Photos(1)

Key Documents

MAB2035

Sigma-Aldrich

Anti-Aggrecan Antibody

CHEMICON®, mouse monoclonal, MK-302

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
MK-302, monoclonal
application:
ELISA
RIA
WB
species reactivity:
canine, sheep, rat, rabbit, mouse, bovine, human
technique(s):
ELISA: suitable
radioimmunoassay: suitable
western blot: suitable
citations:
19

Product Name

Anti-Chondroitin 6 Sulfate Antibody, clone MK-302, ascites fluid, clone MK-302, Chemicon®

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

MK-302, monoclonal

species reactivity

canine, sheep, rat, rabbit, mouse, bovine, human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
radioimmunoassay: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... ACAN(176)

Specificity

Reacts with a wide range of cartilage proteoglycans after either chondroitinase (ABC or AC) or testicular hyaluronidase digestion. Native proteoglycans do not react with MAB2035. Antibody binding to epitope is successfully inhibited by disaccharides of chondroitin-6-sulfate. Recognizes the chondroitin-6-sulfate stubs (preferentially in clusters) of high density cartilage proteoglycans of different species. No cross-reactivity with dermatan sulfate, keratan sulfate or chondroitin-4-sulfate.

Immunogen

Chondroitinase ABC-digested adult human aggrecan.

Application

Anti-Chondroitin 6 Sulfate Antibody, clone MK-302 is an antibody against Chondroitin 6 Sulfate for use in ELISA, RIA & WB.
ELISA at 1:100-1:5,000

RIA at 1:100-1:200.

Chondroitinase ABC digestion prior to antibody reaction is required for antibody reactivity. {0.5U/mL in 100mM Tris-HCL, 30 minutes room temperature} A dramatic proteolytic digestion of the core protein (e.g. with papain or pronase) significantly reduces the antibody reactivity.

Optimal working dilutions must be determined by the end user.

Physical form

Ascites liquid; no preservative.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G D Nicodemus et al.
Journal of biomechanics, 41(7), 1528-1536 (2008-04-18)
Crosslinked poly(ethylene glycol) (PEG) hydrogels are attractive scaffolds for cartilage tissue engineering because of their ability to mimic the aqueous environment and mechanical properties of native cartilage. In this study, hydrogel crosslinking density was varied to study the influence of
Qijing Wu et al.
Cancer immunology research, 10(2), 182-199 (2021-12-23)
Metastatic microsatellite-stable (MSS) colorectal cancer rarely responds to immune checkpoint inhibitors (ICI). Metabolism heterogeneity in the tumor microenvironment (TME) presents obstacles to antitumor immune response. Combining transcriptome (The Cancer Genome Atlas MSS colorectal cancer, n = 383) and digital pathology
Idalis Villanueva et al.
Matrix biology : journal of the International Society for Matrix Biology, 29(1), 51-62 (2009-09-02)
This study aimed to elucidate the role of charge in mediating chondrocyte response to loading by employing synthetic 3D hydrogels. Specifically, neutral poly(ethylene glycol) (PEG) hydrogels were employed where negatively charged chondroitin sulfate (ChS), one of the main extracellular matrix
Robert E Hurst et al.
American journal of physiology. Renal physiology, 310(10), F1074-F1080 (2016-02-26)
Loss of integrity of the protective impermeability barrier in the urothelium has been identified as significant in bladder dysfunction. In this study, we tested the theory that the luminal layer of glycosaminoglycans (GAG) serves as an important component of barrier
Cheng-Fong Chen et al.
International journal of molecular sciences, 22(16) (2021-08-28)
Allogeneic bone grafts are a promising material for bone implantation due to reduced operative trauma, reduced blood loss, and no donor-site morbidity. Although human decellularized allogeneic bone (hDCB) can be used to fill bone defects, the research of revitalizing hDCB

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service