Skip to Content
Merck
All Photos(1)

Key Documents

C7397

Sigma-Aldrich

Cyclophosphamide monohydrate

ISOPAC®

Synonym(s):

2-[Bis(2-chloroethyl)amino]tetrahydro-2H-1,3,2-oxazaphosphorine 2-oxide, Cytoxan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H15Cl2N2O2P · H2O
CAS Number:
Molecular Weight:
279.10
Beilstein:
4678992
EC Number:
MDL number:
UNSPSC Code:
51112507
PubChem Substance ID:
NACRES:
NA.25

Assay

97.0-103.0% (HPLC)

form

powder

mp

49-51 °C (lit.)

storage temp.

2-8°C

SMILES string

[H]O[H].ClCCN(CCCl)P1(=O)NCCCO1

InChI

1S/C7H15Cl2N2O2P.H2O/c8-2-5-11(6-3-9)14(12)10-4-1-7-13-14;/h1-7H2,(H,10,12);1H2

InChI key

PWOQRKCAHTVFLB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Cyclophosphamide, a nitrogen mustard DNA alkylating agent, is used to study its mechanisms of action as an anticancer and antiimmunity drug.

Biochem/physiol Actions

Cyclophosphamide is a cytotoxic nitrogen mustard derivative widely used in cancer chemotherapy. It cross-links DNA, causes strand breakage, and induces mutations. Its clinical activity is associated with a decrease in aldehyde dehydrogenase 1 (ALDH1) activity.
Cytotoxic nitrogen mustard derivative widely used in cancer chemotherapy. It cross-links DNA, causes strand breakage, and induces mutations.

Packaging

Packaged in a 100 mL serum bottle with silicon/PTFE septum and aluminum tear seal.

Caution

Injecting a compatible solvent directly into the vial permits preparation of any desired strength solution without exposure.

Reconstitution

Dissolving the contents in 100 mL of solvent yields a 1% solution.

Legal Information

Isopac is a registered trademark of Merck KGaA, Darmstadt, Germany

ISOPAC of

Product No.
Description
Pricing

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 1B - Muta. 1B - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julia Alicia Boos et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 6(13), 1900294-1900294 (2019-08-06)
The integration of metabolic competence in developmental toxicity assays in vitro is of fundamental importance to better predict adverse drug effects. Here, a microfluidic hanging-drop platform is presented that seamlessly integrates liver metabolism into the embryonic stem cell test (EST).
Monika Kumari et al.
Frontiers in bioengineering and biotechnology, 8, 572077-572077 (2020-12-18)
Currently, amino-terminal PEGylated human granulocyte colony stimulating factor (huG-CSF) is used to prevent and treat neutropenia. Although huG-CSF has been used as a drug for more than 20 years, it has three significant drawbacks: (i) it relies on PEG aldehyde
Leo Luznik et al.
Seminars in oncology, 39(6), 683-693 (2012-12-05)
Allogeneic hematopoietic stem cell transplantation (alloSCT) is a potentially curative therapy for many hematologic and immunologic diseases. Further, partial or full donor hematopoietic chimerism following alloSCT may be sufficient to guarantee immunologic tolerance to solid organs from the same donor
Andrea T Borchers et al.
Autoimmunity reviews, 12(2), 174-194 (2012-09-18)
Lupus nephritis remains one of the most severe manifestations of systemic lupus erythematosus associated with considerable morbidity and mortality. A better understanding of the pathogenesis of lupus nephritis is an important step in identifying more targeted and less toxic therapeutic
Jia Wang et al.
Cell reports, 25(2), 464-477 (2018-10-12)
The urothelium is an epithelia barrier lined by a luminal layer of binucleated, octoploid, superficial cells. Superficial cells are critical for production and transport of uroplakins, a family of proteins that assemble into a waterproof crystalline plaque that helps protect

Articles

Cancer research has revealed that the classical model of carcinogenesis, a three step process consisting of initiation, promotion, and progression, is not complete.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service