Skip to Content
Merck
All Photos(3)

Key Documents

C6013

Sigma-Aldrich

Cholesteryl hemisuccinate tris salt

anionic detergent

Synonym(s):

3β-Hydroxy-5-cholestene 3-hemisuccinate, 5-Cholesten-3β-ol 3-hemisuccinate, CHEMS, CHS, Cholesteryl hydrogen succinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H50O4 · C4H11NO3
CAS Number:
Molecular Weight:
607.86
MDL number:
UNSPSC Code:
12352211
eCl@ss:
39023139
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Quality Level

Assay

≥98% (TLC)

form

powder

functional group

ester

shipped in

ambient

storage temp.

−20°C

SMILES string

NC(CO)(CO)CO.CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@H](CC[C@]4(C)[C@H]3CC[C@]12C)OC(=O)CCC(O)=O

InChI

1S/C31H50O4.C4H11NO3/c1-20(2)7-6-8-21(3)25-11-12-26-24-10-9-22-19-23(35-29(34)14-13-28(32)33)15-17-30(22,4)27(24)16-18-31(25,26)5;5-4(1-6,2-7)3-8/h9,20-21,23-27H,6-8,10-19H2,1-5H3,(H,32,33);6-8H,1-3,5H2/t21-,23+,24+,25-,26+,27+,30+,31-;/m1./s1

InChI key

SLDYONDUXRBLLR-XTCKSVDZSA-N

Application

Cholesteryl hemisuccinate tris salt has been used:
  • to prepare solubilization buffer for glucan synthase assay
  • to prepare immunoprecipitation buffer for immobilization of opioid receptors on paramagnetic beads.
  • as a membrane stabilizer for liposomes and protein synaptogyrin-1
  • as a component of a folate-modified self-microemulsifying drug delivery system.
  • with N-dodecyl β-D-maltoside (DDM) to solubilize erythrocyte ghosts and chemokine receptor CCR1
  • for protein solubilization

Biochem/physiol Actions

Cholesteryl hemisuccinate is an ionizable anionic detergent that stabilizes large unilamellar vesicles. These liposome vesicles act as delivery agents for anticancer drugs, oligonucleotides and antibiotics. Cholesteryl succinate also reportedly exhibits antiproliferative activity and may be clinically useful as antitumor agent.

Preparation Note

Cholesteryl hemisuccinate tris salt yields clear, colorless solution in methanol at 10 mg/ml.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sérgio Simões et al.
Advanced drug delivery reviews, 56(7), 947-965 (2004-04-07)
Strategies used to enhance liposome-mediated drug delivery in vivo include the enhancement of stability and circulation time in the bloodstream, targeting to specific tissues or cells, and facilitation of intracytoplasmic delivery. pH-sensitive liposomes have been developed to mediate the introduction
Cholesteryl hemisuccinate as a membrane stabilizer in dipalmitoylphosphatidylcholine liposomes containing saikosaponin-d.
Ding, W.X., et al.
International Journal of Pharmaceutics, 300, 37-47 (2005)
Megan L Grove et al.
PloS one, 8(7), e68095-e68095 (2013-07-23)
Genotyping arrays are a cost effective approach when typing previously-identified genetic polymorphisms in large numbers of samples. One limitation of genotyping arrays with rare variants (e.g., minor allele frequency [MAF] <0.01) is the difficulty that automated clustering algorithms have to
David Ott et al.
Protein engineering, design & selection : PEDS, 18(3), 153-160 (2005-03-26)
Opioid receptors, like many G protein-coupled receptors (GPCRs), are notoriously unstable in detergents. We have now developed a more stable variant of the mu-opioid receptor (MOR) and also a method for the immobilization of solubilized, functional opioid receptors on a
Samantha J Allen et al.
Protein expression and purification, 66(1), 73-81 (2009-03-12)
Chemokine receptors are a specific class of G-protein-coupled receptors (GPCRs) that control cell migration associated with routine immune surveillance, inflammation and development. In addition to their roles in normal physiology, these receptors and their ligands are involved in a large

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service