Skip to Content
Merck
All Photos(1)

Key Documents

32950

Sigma-Aldrich

4,4′-Diaminodiphenylmethane

≥97.0% (GC)

Synonym(s):

4,4′-Methylenedianiline, MDA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H14N2
CAS Number:
Molecular Weight:
198.26
Beilstein:
474706
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (GC)

mp

88-92 °C

solubility

water: soluble

SMILES string

Nc1ccc(Cc2ccc(N)cc2)cc1

InChI

1S/C13H14N2/c14-12-5-1-10(2-6-12)9-11-3-7-13(15)8-4-11/h1-8H,9,14-15H2

InChI key

YBRVSVVVWCFQMG-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

4,4′-diaminodiphenylmethane acts as a bidentate-bridging ligand, forming hydrogen bonds with its amine hydrogen atoms, and also participates in several non-covalent interactions.

Application

4,4′-Diaminodiphenylmethane was employed:
  • as dummy template for bisphenols imprinting
  • in the synthesis of amino modified multi-walled carbon nanotubes/polydimethylsiloxane
  • as curing agent to study the kinetics of formation of epoxy resin composites
  • in rubber industry as an epoxyresin hardener

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 1B - Muta. 2 - Skin Sens. 1 - STOT RE 2 - STOT SE 1

Target Organs

Liver, Liver,eye - retina

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

429.8 °F - closed cup

Flash Point(C)

221 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bing Yu et al.
Journal of ethnopharmacology, 165, 173-179 (2015-02-25)
Fuzi [the lateral root of Aconitum carmichaeli Debx (Ranunculaceae)] is a well-known traditional medicinal herb used to treat chronic heart failure (CHF). Aconitine-type alkaloids are major alkaloids that are responsible for the pharmacological activity and toxicity of this herb.To investigate
Six transition metal--organic materials with the ditopic 4, 4?-diaminodiphenylmethane ligand: Synthesis, structure characterization and luminescent properties
Chisca D, et al.
Polyhedron, 192, 114844-114844 (2020)
Xiaoli Sun et al.
Journal of chromatography. A, 1343, 33-41 (2014-04-15)
A simple and fast method for both dummy template selection and polymer composition optimization is proposed here. A series of dummy templates for bisphenols imprinting were screened by running them on a non-imprinted polymer (NIP) column with porogen solvent as
Carcinogenic and chronic effects of 4,4'-diaminodiphenylmethane, an epoxyresin hardener.
R Schoental
Nature, 219(5159), 1162-1163 (1968-09-14)
Jessica M Labonté et al.
The ISME journal, 9(11), 2386-2399 (2015-04-08)
Viral infections dynamically alter the composition and metabolic potential of marine microbial communities and the evolutionary trajectories of host populations with resulting feedback on biogeochemical cycles. It is quite possible that all microbial populations in the ocean are impacted by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service