Skip to Content
Merck
All Photos(5)

Key Documents

C6003

Sigma-Aldrich

Chlorophyllin sodium copper salt

commercial grade, powder

Synonym(s):

Chlorophyllin, coppered trisodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H31CuN4Na3O6
CAS Number:
Molecular Weight:
724.15
EC Number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

product name

Chlorophyllin sodium copper salt, commercial grade

grade

commercial grade

form

powder

color

black
green to very dark green

solubility

H2O: 100 mg/mL, dark green

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

SMILES string

[Na+].[Na+].[Na+].CCc1c(C)c2cc3c(C=C)c(C)c4cc5nc(C(CCC([O-])=O)C5C)c(CC([O-])=O)c6c(C([O-])=O)c(C)c(cc1n2)n6[Cu]n34

InChI

1S/C34H36N4O6.Cu.3Na/c1-7-19-15(3)23-12-25-17(5)21(9-10-29(39)40)32(37-25)22(11-30(41)42)33-31(34(43)44)18(6)26(38-33)14-28-20(8-2)16(4)24(36-28)13-27(19)35-23;;;;/h7,12-14,17,21H,1,8-11H2,2-6H3,(H5,35,36,37,38,39,40,41,42,43,44);;;;/q;+2;3*+1/p-5

InChI key

HWDGVJUIHRPKFR-UHFFFAOYSA-I

Looking for similar products? Visit Product Comparison Guide

Application

Chlorophyllin sodium copper salt has been used to study the effect of chlorophyllin against radiation-induced hematopoietic syndrome. It has been used to study the effect of chlorophyllin on the excretion of PhIP (2-amino-1-methyl-6-phenylimidazo[4,5-b]pyridine) and its metabolites into the breast-milk of lactating rats.

Biochem/physiol Actions

Chlorophyllin is a water soluble derivative of chlorophyll. It has chemopreventive properties and forms a non-covalent complex with many mutagenic/carcinogenic molecules. The complex formation reduces absorption or allows excretion of the mutagenic/carcinogenic molecules. Chlorophyllin also exhibits antioxidant, antiapoptotic and immunostimulatory effects. It provides benefits, such as prevention of body odor and increased wound healing.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zivile Luksiene et al.
Journal of photochemistry and photobiology. B, Biology, 105(1), 69-74 (2011-08-03)
The aim of this study was to evaluate efficiency of photosensitization as surface sanitation alternative using model systems when food pathogens, their spores and biofilms were attached to the food-related surface (polyolefine). In addition it was important to compare antibacterial
Distribution and metabolism of 2-amino-1-methyl-6-phenylimidazo[4,5-b]pyridine (PhIP) in female rats and their pups at dietary doses.
Mauthe RJ, et al.
Carcinogenesis, 19, 919-924 (1998)
Wei Li et al.
Nanomedicine (London, England), 7(3), 383-392 (2012-03-06)
How to overcome insufficient drug release is an important issue in the drug-delivery system. Here, a novel temperature and UV dual-control poly(N-isopropylacrylamide [PNIPAM]-co-chlorophyllin) nanogel was prepared via the surfactant-free emulsion polymerization. The introduction of hydrophilic chlorophyllin to the PNIPAM chain
Jun Zhang et al.
The Analyst, 137(18), 4287-4294 (2012-08-02)
The human hepatoma 3B cell line was chosen as an experimental model for in vitro test of drug screening. The drugs included chlorophyllin and its derivatives such as fluo-chlorophyllin, sodium copper chlorophyllin, and sodium iron chlorophyllin. The 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyl-tetrazolium bromide (MTT)
S Wohllebe et al.
Parasitology research, 111(2), 729-733 (2012-04-06)
Tests were conducted to determine the efficiency of the photodynamic substance chlorophyllin to kill different life stages of the protozoan parasite Ichthyophthirius mulftifiliis (Fouquet) which causes the white spot disease in many freshwater fish species. This study has developed a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service