Skip to Content
Merck
All Photos(2)

Documents

C1900

Sigma-Aldrich

Protoporphyrin IX cobalt chloride

Synonym(s):

Cobaltic protoporphyrin IX chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C34H32CoN4O4Cl
CAS Number:
Molecular Weight:
655.03
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.22

form

powder

reaction suitability

core: cobalt
reagent type: catalyst

storage temp.

2-8°C

SMILES string

Cc1c(CCC(O)=O)c2cc3c(CCC(O)=O)c(C)c4cc5nc(cc6c(C=C)c(C)c(cc1n2)n6[Co](Cl)n34)c(C)c5C=C

InChI

1S/C34H34N4O4.ClH.Co/c1-7-21-17(3)25-13-26-19(5)23(9-11-33(39)40)31(37-26)16-32-24(10-12-34(41)42)20(6)28(38-32)15-30-22(8-2)18(4)27(36-30)14-29(21)35-25;;/h7-8,13-16H,1-2,9-12H2,3-6H3,(H4,35,36,37,38,39,40,41,42);1H;/q;;+3/p-3/b25-13-,26-13-,27-14-,28-15-,29-14-,30-15-,31-16-,32-16-;;

InChI key

WWNXKQFLDNSNDN-HXFTUNQESA-K

Application

Reactant involved in studies of:
  • Enzyme models of hCBS heme group function
  • Electrocatalytic reduction of oxygen at electropolymerized films of porphyrins
  • Electrochemical redox behavior
  • Nitrogen(II) oxide coordination by metal porphyrins

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sheng Chen et al.
Frontiers in cell and developmental biology, 8, 127-127 (2020-03-21)
In vertebrate, the nucleus pulposus (NP), which is an essential component of the intervertebral disk, is constantly impacted by fluid shear stress (FSS); however, molecular mechanism(s) through which FSS modulates the NP homeostasis is poorly understood. Here we show that
Ye Tao et al.
Investigative ophthalmology & visual science, 60(15), 5080-5094 (2019-12-12)
Retinitis pigmentosa (RP) causes progressive photoreceptor degeneration in the retina. The N-methyl-N-nitrosourea (MNU)-administered mouse is used as a chemically induced RP model with rapid progression rate. This study was designed to study heme oxygenase-1 (HO-1) expression in the MNU-administered mice
Yu Wang et al.
Aging, 12(13), 12869-12895 (2020-07-01)
Differences in microRNA (miRNA) expression after intracerebral hemorrhage (ICH) have been reported in human and animal models, and miRNAs are being investigated as a new treatment for inflammation and oxidative stress after ICH. In this study, we found that microRNA-183-5p
Shuang Li et al.
Antioxidants & redox signaling, 30(13), 1635-1650 (2018-08-08)
The risk factors promoting acute kidney injury (AKI) to chronic kidney disease (CKD) progression remain largely unknown. The aim of the present study was to investigate whether hyperhomocysteinemia (Hhcy) accelerates the development of renal fibrosis after AKI. Hhcy aggravated ischemia-reperfusion-induced

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service