Skip to Content
Merck
All Photos(1)

Documents

C5405

Sigma-Aldrich

Chicken Serum

USA origin, sterile-filtered, suitable for cell culture

Synonym(s):

chicken sera

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

biological source

chicken

sterility

sterile-filtered

form

liquid

composition

hemoglobin, ≤60 mg/dL

origin

USA origin

technique(s)

cell culture | mammalian: suitable

impurities

≤50 EU/mL endotoxin

shipped in

dry ice

storage temp.

−20°C

Application

Chicken Serum has been used:
  • for pellet resuspension following centrifugation, to prepare zymosan suspension for respiratory burst assay
  • as a supplement to Dulbecco′s modified Eagle′s medium (DMEM) to culture HeLa P cells
  • for blocking in immunofluorescence (IF) double-staining

Chicken serum is used in parasitology to study excystation, growth, survival and egg production of the microphallid trematode Maritrema novaezealandensis. Chicken serum is used for the serial cultivation of chicken keratinocytes.

Biochem/physiol Actions

Animal sera serve as a cost-effective alternative to basal cell culture media supplement. They are commonly used in immunodetection protocols such as immunohistochemistry (IHC), immunocytochemistry (ICC), and western blot to block nonspecific binding. Chicken serum is suitable for use in parasitology to study excystation, growth, survival, and egg production of the microphallid trematode Maritrema novaezealandensis. It is used for the serial cultivation of chicken keratinocytes.

Analysis Note

Endotoxin tested

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Géraldine Rouquet et al.
Journal of bacteriology, 191(13), 4427-4440 (2009-04-21)
We identified a carbohydrate metabolic operon (frz) that is highly associated with extraintestinal pathogenic Escherichia coli (ExPEC) strains. The frz operon codes for three subunits of a phosphoenolpyruvate:carbohydrate phosphotransferase system (PTS) transporter of the fructose subfamily, for a transcriptional activator
B L Fredensborg et al.
Parasitology research, 95(5), 310-313 (2005-02-01)
In this study, the effect of the composition of culture medium on excystation, growth, survival and egg production was investigated for the recently discovered microphallid trematode Maritrema novaezealandensis. Metacercariae from the two second intermediate crab hosts, Macrophthalmus hirtipes and Halicarcinus
Paula A Bousquet et al.
Biochemical and biophysical research communications, 495(1), 1562-1566 (2017-12-03)
Immunotherapy is a growing field in cancer research. A privileged tumor-associated antigen that has received much attention is N-glycolyl (NeuGc) GM3. This ganglioside is present in several types of cancer, but is almost undetectable in human healthy tissues. However, its
Jeong Hyo Lee et al.
BMC molecular and cell biology, 21(1), 40-40 (2020-05-31)
In the poultry and livestock industries, precise genetic information is crucial for improving economic traits. Thus, functional genomic studies help to generate faster, healthier, and more efficient animal production. Chicken myoblast cells, which are required for muscle development and regeneration
Tom W Anderson et al.
Molecular biology of the cell, 19(11), 5006-5018 (2008-09-19)
In migrating fibroblasts actomyosin II bundles are graded polarity (GP) bundles, a distinct organization to stress fibers. GP bundles are important for powering cell migration, yet have an unknown mechanism of formation. Electron microscopy and the fate of photobleached marks

Articles

Find answers to frequently asked questions (FAQs) about FBS and other serum products, and links to related products.

Find answers to frequently asked questions (FAQs) about FBS and other serum products, and links to related products.

Find answers to frequently asked questions (FAQs) about FBS and other serum products, and links to related products.

Find answers to frequently asked questions (FAQs) about FBS and other serum products, and links to related products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service