Skip to Content
Merck
All Photos(1)

Key Documents

700122P

Avanti

campestanol

Avanti Research - A Croda Brand 700122P, powder

Synonym(s):

5α-campestan-3β-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H50O
CAS Number:
Molecular Weight:
402.70
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (700122P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 700122P

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@@H]1C[C@H]2[C@@]([C@@H]3[C@H]([C@H]4[C@@]([C@H](CC4)[C@@H](CC[C@H](C(C)C)C)C)(CC3)C)CC2)(CC1)C

InChI

1S/C28H50O/c1-18(2)19(3)7-8-20(4)24-11-12-25-23-10-9-21-17-22(29)13-15-27(21,5)26(23)14-16-28(24,25)6/h18-26,29H,7-17H2,1-6H3/t19-,20-,21+,22+,23+,24-,25+,26+,27+,28-/m1/s1

InChI key

ARYTXMNEANMLMU-ATEDBJNTSA-N

General description

Campestanol has a methyl group at the C-24. It comprises the 10-30% of stanol content in vegetable oil. It is naturally occurring and has similar structure as cholesterol. Campestanol is synthesized by the hydrogenation of campesterol.
Campestanol is a saturated plant stanol.

Biochem/physiol Actions

Campestanol absorption in intestine is similar to that of cholesterol absorption pathway. However, in sitosterolemic homozygotes, it is hyperabsorbed from the intestine.
Intestinal absorption of campestanol is lesser compared to cholesterol due to the presence of side chain. Due to the structural similarity with cholesterol, campestanol reduces cholesterol absorption in the intestine and the plasma level of LDL cholesterol.

Packaging

5 mL Amber Glass Screw Cap Vial (700122P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Campestanol (24-methyl-5alpha-cholestan-3beta-ol) absorption and distribution in New Zealand white rabbits: effect of dietary sitostanol
Xu G, et al.
Metabolism, Clinical and Experimental, 48(3), 363-368 (1999)
SOFW Journal, 149-149 (2002)
Campestanol (24-methyl-5alpha-cholestan-beta-ol) absorption and distribution in New Zealand white rabbits: effect of dietary sitostanol
Xu G, et al.
Metabolism and Disease, 48(3), 363-368 (1999)
Hyperabsorption and retention of campestanol in a sitosterolemic homozygote: comparison with her mother and three control subjects
Salen, Gerald and Xu, Guorong and Tint, GS and Batta, AK and Shefer, Sarah
Journal of Lipid Research, 41(11), 1883-1889 (2000)
Enhancing Extraction Processes in the Food Industry (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service