Skip to Content
Merck
All Photos(4)

Key Documents

C5132

Sigma-Aldrich

Carbazole

≥95% purity (GC), powder

Synonym(s):

Dibenzopyrrole; Diphenylenimine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H9N
CAS Number:
Molecular Weight:
167.21
Beilstein:
3956
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47
Pricing and availability is not currently available.

Product Name

Carbazole, ≥95% (GC)

vapor pressure

400 mmHg ( 323 °C)

Assay

≥95% (GC)

form

powder

technique(s)

titration: suitable

color

white to tan

bp

355 °C (lit.)

mp

243-246 °C (lit.)

solubility

acetone: 50 mg/mL

density

1.1 g/cm3 at 18 °C

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

c1ccc2c(c1)[nH]c3ccccc23

InChI

1S/C12H9N/c1-3-7-11-9(5-1)10-6-2-4-8-12(10)13-11/h1-8,13H

InChI key

UJOBWOGCFQCDNV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Carbazole is an aromatic heterocyclic organic molecule.

Application

Carbazole has been used in the uronic acid assay.[1]

Biochem/physiol Actions

Carbazole mainly interacts with DNA and damages it. These events suppress the synthesis of new DNA or RNA. Carbazole derivatives are involved in antimicrobial, antitumor, antiepileptic, antihistaminic, antioxidant, anti-inflammatory, antidiarrhoeal, analgesic, neuroprotective, and pancreatic lipase suppression activities.

Quality

May contain dark particles.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Carc. 2 - Muta. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

428.0 °F - closed cup

Flash Point(C)

220.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A complete set of hyaluronan fragments obtained from hydrolysis catalyzed by hyaluronidase: Application to studies of hyaluronan mass distribution by simple HPLC devices.
Tranchepain F, et al.
Analytical Biochemistry, 348, 232-242 (2006)
CARBAZOLE DERIVATIVES IN CANCER TREATMENT-A REVIEW.
Kanneti Naga M, et al.
World Journal of Pharmacy and Pharmaceutical Sciences (2015)
Shen H Tan et al.
Organic letters, 14(22), 5621-5623 (2012-10-31)
The racemic modification of the Aspidosperma alkaloid limaspermidine (1) has been prepared in ten steps including one involving a Raney-cobalt-mediated tandem reductive cyclization of nitrile 8 to give the tetracyclic system 9b. Compound (±)-1 has been converted over two steps
Jong-Kwan Bin et al.
Advanced materials (Deerfield Beach, Fla.), 24(21), 2911-2915 (2012-05-03)
A new carbazole derivative with two carbazole moieties on the C3 and C6 positions of carbazole and triphenylsilane directly linked to the N of carbazole is successfully used as a highly efficient blue phosphorescent host in an organic light-emitting diode
Yuji Ashikawa et al.
BMC structural biology, 12, 15-15 (2012-06-26)
Dihydroxylation of tandemly linked aromatic carbons in a cis-configuration, catalyzed by multicomponent oxygenase systems known as Rieske nonheme iron oxygenase systems (ROs), often constitute the initial step of aerobic degradation pathways for various aromatic compounds. Because such RO reactions inherently

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service