Skip to Content
Merck
All Photos(1)

Key Documents

46100

Supelco

Carbazole

VETRANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H9N
CAS Number:
Molecular Weight:
167.21
Beilstein:
3956
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor pressure

400 mmHg ( 323 °C)

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

355 °C (lit.)

mp

243-246 °C (lit.)

application(s)

clinical
environmental

format

neat

SMILES string

c1ccc2c(c1)[nH]c3ccccc23

InChI

1S/C12H9N/c1-3-7-11-9(5-1)10-6-2-4-8-12(10)13-11/h1-8,13H

InChI key

UJOBWOGCFQCDNV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Carc. 2 - Muta. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

428.0 °F - closed cup

Flash Point(C)

220.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuji Ashikawa et al.
BMC structural biology, 12, 15-15 (2012-06-26)
Dihydroxylation of tandemly linked aromatic carbons in a cis-configuration, catalyzed by multicomponent oxygenase systems known as Rieske nonheme iron oxygenase systems (ROs), often constitute the initial step of aerobic degradation pathways for various aromatic compounds. Because such RO reactions inherently
Guodong Ji et al.
Journal of hazardous materials, 225-226, 1-7 (2012-05-23)
We examined the effects of power and treatment time on the ultrasonically enhanced ozonation of carbazole dissolved in APG(1214) surfactant solutions, including an analysis of the mechanism of OH radical formation, the zeta potential of the colloidal suspension, the influence
Jae-Min Suk et al.
Organic letters, 14(19), 5018-5021 (2012-09-18)
Chiral organic anions such as camphorsulfonates and cAMP give rise to the preferential formation of a one-handed helix of an indolocarbazole foldamer, thus inducing characteristic circular dichroic (CD) signals. Moreover, the on and off switching of the chiroptical signal can
Xiaofeng Liu et al.
Advanced materials (Deerfield Beach, Fla.), 24(33), 4505-4510 (2012-06-22)
An arylated-carbazole conjugated polymer with a deep HOMO level has been developed. Solar cells based on blends of PCX3 and PC(71) BM show efficiency of 3.9% with a V(oc) of 0.96 V. The device performance can be improved to 5.1%
Shen H Tan et al.
Organic letters, 14(22), 5621-5623 (2012-10-31)
The racemic modification of the Aspidosperma alkaloid limaspermidine (1) has been prepared in ten steps including one involving a Raney-cobalt-mediated tandem reductive cyclization of nitrile 8 to give the tetracyclic system 9b. Compound (±)-1 has been converted over two steps

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service