Skip to Content
Merck
All Photos(2)

Key Documents

C1988

Sigma-Aldrich

Cycloheximide

Biotechnology Performance Certified

Synonym(s):

3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide, Actidione, Naramycin A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H23NO4
CAS Number:
Molecular Weight:
281.35
Beilstein:
88868
EC Number:
MDL number:
UNSPSC Code:
51102829
PubChem Substance ID:
NACRES:
NA.85
Pricing and availability is not currently available.

grade

Biotechnology Performance Certified

Quality Level

form

powder

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

solubility

ethanol: 50 mg/mL

antibiotic activity spectrum

fungi
yeast

Mode of action

protein synthesis | interferes

storage temp.

2-8°C

SMILES string

[H][C@]1(C[C@@H](C)C[C@H](C)C1=O)[C@H](O)CC2CC(=O)NC(=O)C2

InChI

1S/C15H23NO4/c1-8-3-9(2)15(20)11(4-8)12(17)5-10-6-13(18)16-14(19)7-10/h8-12,17H,3-7H2,1-2H3,(H,16,18,19)/t8-,9-,11-,12+/m0/s1

InChI key

YPHMISFOHDHNIV-FSZOTQKASA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Cycloheximide is widely used for selection of CHX-resistant strains of yeast and fungi, controlled inhibition of protein synthesis for detection of short-lived proteins and super-induction of protein expression, and apoptosis induction or facilitation of apoptosis induction by death receptors. It has been shown to selectively clear macrophages in atherosclerotic plaques[1] and activate cumulus-free equine oocytes.[2]

Biochem/physiol Actions

Cycloheximide (CHX) is an antibiotic produced by S. griseus that inhibits protein biosynthesis in eukaryotes. It inactivate transferase II enzyme that is involved in peptide chain elongation.[3] More recent studies revealed that CHX binds the 60S ribosome and specifically prevents the translocation step in elongation.[4]
Mode of Action: Translation inhibition in eukaryotes resulting in cell growth arrest and cell death. CHX is widely used for the selection of CHX-resistant strains of yeast and fungi, controlled inhibition of protein synthesis for detection of short-lived proteins and super-induction of protein expression, and apoptosis induction or facilitation of apoptosis induction by death receptors.

Activity Spectrum: Active against yeast and fungi like Candida, Aspergillus, Saccharomyces, Penicillium

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Keep in a dry place.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Chronic 2 - Muta. 2 - Repr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Valerie Croons et al.
The Journal of pharmacology and experimental therapeutics, 320(3), 986-993 (2006-12-01)
Macrophages are an essential component of unstable atherosclerotic plaques and play a pivotal role in the destabilization process. We have demonstrated previously that local delivery of the mammalian target of rapamycin (mTOR) inhibitor everolimus selectively clears macrophages in rabbit plaques.
Mechanism of cycloheximide inhibition of protein synthesis in a cell-free system prepared from rat liver.
B S Baliga et al.
The Journal of biological chemistry, 244(16), 4480-4489 (1969-08-25)
Tilman Schneider-Poetsch et al.
Nature chemical biology, 6(3), 209-217 (2010-02-02)
Although the protein synthesis inhibitor cycloheximide (CHX) has been known for decades, its precise mechanism of action remains incompletely understood. The glutarimide portion of CHX is seen in a family of structurally related natural products including migrastatin, isomigrastatin and lactimidomycin
Y H Choi et al.
Reproduction (Cambridge, England), 122(1), 177-183 (2001-06-27)
Two different culture media (TCM-199 and follicular fluid), two activation treatments (10 and 50 micromol calcium ionophore l(-1)) and three culture periods with cycloheximide were evaluated to find effective culture conditions for activation of cumulus-free equine oocytes. Oocytes were collected
Nina Xue et al.
Cellular and molecular life sciences : CMLS, 76(17), 3433-3447 (2019-04-14)
Enhancement of insulin-like growth factor 1 receptor (IGF-IR) degradation by heat shock protein 90 (HSP90) inhibitor is a potential antitumor therapeutic strategy. However, very little is known about how IGF-IR protein levels are degraded by HSP90 inhibitors in pancreatic cancer

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service