Skip to Content
Merck
All Photos(1)

Key Documents

75051

Supelco

Lycopene

analytical standard

Synonym(s):

E160d, ψ,ψ-Carotene, 2,6,10,14,19,23,27,31-Octamethyl-dotriaconta-2,6,8,10,12,14,16,18,20,22,24,26,30-tridecaene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H56
CAS Number:
Molecular Weight:
536.87
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥85.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

shipped in

dry ice

storage temp.

-65 to -96°C

SMILES string

C\C(C)=C\CC\C(C)=C\C=C\C(C)=C\C=C\C(C)=C\C=C\C=C(C)\C=C\C=C(C)\C=C\C=C(/C)CC\C=C(\C)C

InChI

1S/C40H56/c1-33(2)19-13-23-37(7)27-17-31-39(9)29-15-25-35(5)21-11-12-22-36(6)26-16-30-40(10)32-18-28-38(8)24-14-20-34(3)4/h11-12,15-22,25-32H,13-14,23-24H2,1-10H3/b12-11+,25-15+,26-16+,31-17+,32-18+,35-21+,36-22+,37-27+,38-28+,39-29+,40-30+

InChI key

OAIJSZIZWZSQBC-GYZMGTAESA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lycopene is a natural compound that belongs to the group of non-provitamin carotenoids and is responsible for the red color of tomatoes and other red-colored fruits and vegetables. It shows a stronger anti-oxidant activity compared to the other carotenoids.

Application

This analytical standard can also be used as follows:
  • Development of a Near-infrared spectroscopic (NIRS) technique for the quantitative analysis of lycopene in dehydrated tomato samples
  • Multi-residue analysis of thiamine, riboflavin, niacin, pyridoxine, folic acid, ascorbic acid, retinol, γ-tocopherol, phylloquinone, lycopene, and β-carotene in cherry tomato and pharmaceutical formulation samples using liquid chromatography (LC) combined with UV detection
  • Quantitative analysis of nine carotenoids in dietary supplement samples by an ultra-high performance supercritical fluid chromatography-photodiode array detection (UHPSFC-PDA) based method
  • Simultaneous determination of lycopene and ubidecarenone in their combined pharmaceutical dosage form by reversed phase-high performance liquid chromatography (RP-HPLC) coupled with UV detection at 400 nm, in accordance with International Conference on Harmonization ICH Q2 R guidelines
  • Raman spectroscopic analysis of orange-fleshed sweet potato samples to measure their carotenoid content
  • Simultaneous measurement of chlorophylls (a & b) and carotenoids (β-carotene and lycopene) in fruit samples of strawberry, apricot, and raspberry by two UV-vis spectrophotometric methods

Biochem/physiol Actions

Antioxidant micronutrient of tomatoes associated with decreased risk for cancer and cardiovascular disease. Enhances gap juction communication between cells via upregulation of connexin 43 and reduces proliferation of cancer cells in culture. Inhibits cholesterol synthesis and enhances low-density lipoprotein degradation.

Other Notes

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sebastian Dams et al.
International journal of food sciences and nutrition, 71(6), 769-780 (2020-02-18)
The major aim of this controlled, randomised, open-labelled, parallel-grouped, clinical trial was to investigate whether supplementation with different dosages of omega-3 fatty acids (0.5 g/d and 1 g/d) from a plant-based fatty acid supplement affected omega-3-indices (O3I) in well-nourished, healthy people. In
Sebastian Dams et al.
International journal for vitamin and nutrition research. Internationale Zeitschrift fur Vitamin- und Ernahrungsforschung. Journal international de vitaminologie et de nutrition, 91(1-2), 77-86 (2019-11-16)
Vitamins and carotenoids are organic compounds that are important for vital functions of the human organism. Since the human body is not able to synthesize most of these micronutrients, they need to be supplied by the intake of food or
Jinhyuk Kim et al.
Foods (Basel, Switzerland), 9(8) (2020-07-28)
The chemical stability of the lipophilic bioactives encapsulated in emulsions can be influenced by emulsion droplet interfacial characteristics as well as by the ability of antioxidants incorporated in emulsion to prevent the degradation of the encapsulated compounds. Therefore, this study
Edward M Barbieri et al.
Cell, 171(6), 1453-1467 (2017-11-21)
We describe a multiplex genome engineering technology in Saccharomyces cerevisiae based on annealing synthetic oligonucleotides at the lagging strand of DNA replication. The mechanism is independent of Rad51-directed homologous recombination and avoids the creation of double-strand DNA breaks, enabling precise
L Minutoli et al.
Current medicinal chemistry, 20(10), 1306-1312 (2013-02-26)
Benign prostatic hyperplasia (BPH) is a major health concern that is likely to have an increasing impact in line with the gradual aging of the population. BPH is characterized by smooth muscle and epithelial proliferation primarily within the prostatic transition

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service