Skip to Content
Merck
All Photos(1)

Key Documents

C4486

Sigma-Aldrich

Bovine Collagen Type II

from bovine articular cartilage, liquid, 1 mg/mL, FITC conjugated, suitable for collagenase activity assay

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32
Pricing and availability is not currently available.

Product Name

Collagen Type II−FITC Conjugate from bovine articular cartilage, ~1 mg/mL, solution

Assay

≥95% (HPLC)

Quality Level

form

solution

concentration

~1 mg/mL

shipped in

wet ice

storage temp.

−20°C

Related Categories

Application

Substrate for assaying mammalian collagenase activity in cell culture.

Physical form

Supplied in 0.01 M acetic acid.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eric K Patterson et al.
Pathophysiology : the official journal of the International Society for Pathophysiology, 28(3), 320-327 (2022-04-03)
A full understanding of the molecular mechanisms implicated in the etiopathogenesis of juvenile idiopathic arthritis (JIA) is lacking. A critical role for leukocyte proteolytic activity (e.g., elastase and cathepsin G) has been proposed. While leukocyte elastase's (HLE) role has been
I J Anderson et al.
American journal of human genetics, 46(5), 896-901 (1990-05-01)
Spondyloepiphyseal dysplasia congenita (SEDC) is an autosomal dominantly inherited chondrodysplasia characterized by disproportionate short stature (short trunk), abnormal epiphyses, and flattened vertebral bodies. Manifestations are present at birth. We ascertained a 4-generation family exhibiting the clinical manifestations of the disorder.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service