Skip to Content
Merck
All Photos(2)

Key Documents

C9301

Sigma-Aldrich

Chicken Collagen Type II

from chicken sternal cartilage, powder, suitable for cell culture

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.75

Product Name

Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture

biological source

chicken (Sternal cartilage)

Quality Level

type

Type II (Miller)

product line

BioReagent

form

powder

packaging

glass bottle of 100 mg
poly bottle of 25 mg
glass bottle of 5 mg

concentration

60-80% (biuret)

technique(s)

cell culture | mammalian: suitable

surface coverage

6‑10 μg/cm2

solubility

acetic acid: 0.5-2.0 mg/mL (Dissolve for several hours at 2-8 °C, occasionally swirling.)

UniProt accession no.

Binding Specificity

Peptide Source: Fibrinogen

shipped in

wet ice

storage temp.

2-8°C

Gene Information

chicken ... COL2A1(395069)

General description

Collagen is classified into a number of structurally and genetically distinct types. We use the nomenclature proposed by Bornstein and Traub. Do not confuse Sigma type designations with recognized collagen classification types.
Collagen is made of three polypeptide chains. Type 2 collagen is ubiquitously present in the hyaline cartilage. It is a homotrimer made of three α1 chains. Type 2 collagen is also predominant in nucleus pulposus of the vertebral disc and vitreous body of the eye. The proteoglycan obtained from the chicken sternal cartilage is similar to type nine collagen structure.

Application

Collagen from chicken sternal cartilage has been used:

  • in enzyme–linked immunosorbent assay (ELISA).
  • chondrocyte-mediated tissue production in vitro.
  • induction, treatment, and assessment of collagenα induced arthritis (CIA).
  • cell proliferation assay.
  • as a coating for cell culture surfaces.

Biochem/physiol Actions

Collagen breaks down metabolically in the body to release N-telopeptide, which is the N-terminus of collagen. There is also C-telopeptide, which is presumably the C-terminus. N-telopeptide is released in urine, and its detection in diagnostic tests is used to screen for osteoporosis.

Features and Benefits

This collagen has been tested in culture with mammalian cells to verify it is low in endotoxin content.

Other Notes

Major component of articular cartilage.

Preparation Note

Prepared by a modification of the method of Trentham, D.E., et al.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clinical Impact of Bone and Connective Tissue Markers (2012)
Lipopolysaccharide Increases the Incidence of Collagen?Induced Arthritis in Mice Through Induction of Protease HTRA?1 Expression.
Hou Y, et al.
Arthritis and Rheumatism, 65(11), 2835-2846 (2013)
Biology of Proteoglycans (2013)
Shifeng Pan et al.
Experimental and therapeutic medicine, 23(2), 139-139 (2022-01-25)
Rheumatoid arthritis (RA) is a chronic autoimmune disease that causes erosion of articular cartilage and bone and has adverse effects on both patients and livestock animals. The aim of the present study was to investigate the role of interleukin-1β (IL-1β)
Mesenchymal-Epithelial Interactions in Neural Development (2013)

Articles

Cancer stem cell media, spheroid plates and cancer stem cell markers to culture and characterize CSC populations.

Cancer stem cell media, spheroid plates and cancer stem cell markers to culture and characterize CSC populations.

Cancer stem cell media, spheroid plates and cancer stem cell markers to culture and characterize CSC populations.

Cancer stem cell media, spheroid plates and cancer stem cell markers to culture and characterize CSC populations.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service