Skip to Content
Merck
All Photos(1)

Key Documents

C4369

Sigma-Aldrich

Calcipotriol hydrate

≥98% (HPLC), powder, Vitamin D3 analog

Synonym(s):

(5Z,7E,22E,24S)-24-Cyclopropyl-9,10-secochola-5,7,10(19),22-tetraene-1alpha,3beta,24-triol, Calcipotriene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H40O3 · xH2O
CAS Number:
Molecular Weight:
412.60 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

Calcipotriol hydrate, ≥98% (HPLC)

Quality Level

Assay

≥98% (HPLC)

form

powder

solubility

DMSO: 15 mg/mL, clear

storage temp.

room temp

SMILES string

O.C[C@H](\C=C\[C@@H](O)C1CC1)C2CCC3\C(CCC[C@]23C)=C\C=C4\C[C@@H](O)C[C@H](O)C4=C

InChI

1S/C27H40O3.H2O/c1-17(6-13-25(29)20-8-9-20)23-11-12-24-19(5-4-14-27(23,24)3)7-10-21-15-22(28)16-26(30)18(21)2;/h6-7,10,13,17,20,22-26,28-30H,2,4-5,8-9,11-12,14-16H2,1,3H3;1H2/b13-6+,19-7+,21-10-;/t17-,22-,23-,24+,25-,26+,27-;/m1./s1

InChI key

XBKHACNRWFKJNC-MANNPBRJSA-N

Biochem/physiol Actions

Calcipotriol is a synthetic derivative of calcitriol, or Vitamin D.
Calcipotriol, a synthetic derivative of calcitriol or Vitamin D, is used in the treatment of psoriasis and marketed under the trade name Dovonex. It has comparable affinity with calcitriol (Vit. D) for the Vitamin D receptor (VDR), while being less than 1% as active as the calcitriol in regulating calcium metabolism. VDR belongs to the steroid/thyroid receptor superfamily, and is found on the cells of many different tissues including the thyroid, bone, kindney, and T cells of the immune system. Binding of calcipotriol to the VDR modulates the T cells gene transcription of cell differentiation and proliferation-related genes.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Oral - Eye Irrit. 2 - Repr. 2 - Skin Sens. 1B - STOT RE 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Sato-Deguchi et al.
The British journal of dermatology, 167(1), 77-84 (2012-03-06)
Psoriasis is a chronic inflammatory skin disease of unknown aetiology, and an active form of vitamin D(3) (1α,25-dihydroxyvitamin D(3)) and its analogues (VD3As) are widely used topical reagents for psoriasis treatment. Besides their well-known calcium homeostasis functions, VD3As have been
Andrés Tirado-Sánchez et al.
Cutis, 90(3), 140-144 (2012-10-26)
Calcipotriene ointment is widely used in the topical treatment of psoriasis, with tacrolimus ointment as an effective alternative in controlling stable plaque psoriasis. The efficacy of the combination of both products on stable plaque psoriasis has not been assessed in
Steven R Feldman et al.
American journal of clinical dermatology, 13(4), 261-271 (2012-05-17)
Topical calcipotriene is frequently prescribed for the treatment of plaque-type psoriasis. Calcipotriene is currently available in the US as an ointment, a solution, a cream, and in a fixed-dose combination ointment with betamethasone dipropionate. Calcipotriene 0.005% has recently been formulated
J Meephansan et al.
Clinical and experimental dermatology, 37(8), 889-896 (2012-08-29)
Vitamin D3 is a potent regulator of cell growth, differentiation and death, tumour invasion, and angiogenesis. Production of matrix metalloproteinase (MMP)-9 and MMP-13 by tumour cells may promote tumour growth, invasion and metastasis. To investigate whether calcipotriol could suppress the
Zuzana Hegyi et al.
The Journal of investigative dermatology, 132(5), 1416-1424 (2012-03-10)
The antimicrobial peptides (AMP) psoriasin (S100A7) and koebnerisin (S100A15) are differently induced in psoriatic skin. They act synergistically as chemoattractants and "alarmins" to amplify inflammation in psoriasis. Th17 cytokines are key players in psoriasis pathogenesis and vitamin D analogs feature

Articles

We offer many products related to non-steroid nuclear receptors for your research needs.

We offer many products related to non-steroid nuclear receptors for your research needs.

We offer many products related to non-steroid nuclear receptors for your research needs.

We offer many products related to non-steroid nuclear receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service