Skip to Content
Merck
All Photos(1)

Key Documents

679102

Millipore

Vitamin D3, 25-Hydroxy-

Metabolite of vitamin D3 via a single cytochrome P450-mediated hydroxylation.

Synonym(s):

Vitamin D3, 25-Hydroxy-, 25-OH-D₃

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H44O2
CAS Number:
Molecular Weight:
400.64
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.21
Pricing and availability is not currently available.

Quality Level

Assay

≥99% (LC/MS)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

ethanol: 10 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

O[C@H]1CCC(=C)\C(=C/C=C2/[C@H]3[C@@]([C@H](CC3)[C@@H](CCCC(O)(C)C)C)(CCC/2)C)\C1

InChI

1S/C27H44O2/c1-19-10-13-23(28)18-22(19)12-11-21-9-7-17-27(5)24(14-15-25(21)27)20(2)8-6-16-26(3,4)29/h11-12,20,23-25,28-29H,1,6-10,13-18H2,2-5H3/b21-11+,22-12-/t20-,23+,24-,25+,27-/m1/s1

InChI key

JWUBBDSIWDLEOM-DTOXIADCSA-N

General description

Metabolite of vitamin D3 via a single cytochrome P450-mediated hydroxylation. Further metabolized to 1α, 25-dihydroxy vitamin D by a second cytochrome P450-mediated hydroxylation. A major circulating metabolite of vitamin D3.

Packaging

Packaged under inert gas

Warning

Toxicity: Toxic & Carcinogenic / Teratogenic (G)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 4 months at -20°C.

Other Notes

DeLuca, H.F., and Schnoes, H.K. 1983. Annu. Rev. Biochem.52, 411.
Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - STOT RE 1 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service