Skip to Content
Merck
All Photos(1)

Key Documents

C4032

Sigma-Aldrich

Casein, Dephosphorylated from bovine milk

lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.61

biological source

bovine milk

Assay

≥85% (biuret)

form

lyophilized powder

UniProt accession no.

storage temp.

2-8°C

SMILES string

[P](=O)(OCC(NC(=O)C(NC(=O)C(N)Cc1ccccc1)CCC(=O)N)C(=O)NC(CCC(=O)O)C(=O)NC(CCC(=O)O)C(=O)NC(CCC(=O)N)C(=O)NC(CCC(=O)N)C(=O)NC(CCC(=O)N)C(=O)NC(C(O)C)C(=O)NC(CCC(=O)O)C(=O)NC(CC(=O)O)C(=O)NC(CCC(=O)O)C(=O)NC(CC(C)C)C(=O)NC(CCC(=O)N)C(=O)NC(CC(=O)O)C(=O)NC(C

InChI

1S/C81H125N22O39P/c1-36(2)31-50(76(132)94-43(15-24-57(87)108)71(127)101-52(34-64(120)121)78(134)98-49(81(137)138)11-7-8-30-82)99-72(128)47(19-28-61(114)115)95-77(133)51(33-63(118)119)100-73(129)48(20-29-62(116)117)97-80(136)65(37(3)104)103-75(131)44(16-25-58(88)109)92-68(124)42(14-23-56(86)107)90-67(123)41(13-22-55(85)106)91-69(125)45(17-26-59(110)111)93-70(126)46(18-27-60(112)113)96-79(135)53(35-142-143(139,140)141)102-74(130)40(12-21-54(84)105)89-66(122)39(83)32-38-9-5-4-6-10-38/h4-6,9-10,36-37,39-53,65,104H,7-8,11-35,82-83H2,1-3H3,(H2,84,105)(H2,85,106)(H2,86,107)(H2,87,108)(H2,88,109)(H,89,122)(H,90,123)(H,91,125)(H,92,124)(H,93,126)(H,94,132)(H,95,133)(H,96,135)(H,97,136)(H,98,134)(H,99,128)(H,100,129)(H,101,127)(H,102,130)(H,103,131)(H,110,111)(H,112,113)(H,114,115)(H,116,117)(H,118,119)(H,120,121)(H,137,138)(H2,139,140,141)

InChI key

BECPQYXYKAMYBN-UHFFFAOYSA-N

Gene Information

General description

Dephosphorylated casein is prepared by the action of potato acid phosphatase. It displays improved solubility and less foaming.

Application

Casein, Dephosphorylated from bovine milk has been used as a substrate in in vitro kinase assays of Arabidopsis thaliana GSK3/Shaggy like kinase, casein kinase II and NIMA (never in mitosis A)-related kinase 2 (Nek2).

Biochem/physiol Actions

Clusters of phosphoserine in casein chelate iron and reduce its bioavailability. Partial enzymatic digestion of casein unmasks phosphorylated residues to alkaline phosphatase, permitting dephosphorylation and increasing the bioavailability of iron.
Dephosphorylated casein lacks a net negative charge and is less sensitive to calcium. It is useful in food industry.

Quality

At least 70% dephosphorylated.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Functional Properties of Dephosphorylated Bovine Whole Casein1
Van Hekken DL and Strange, ED
Journal of Dairy Science, 76(11) (1993)
Wei Liu et al.
The Plant journal : for cell and molecular biology, 36(2), 189-202 (2003-10-11)
Casein kinases are critical in cell division and differentiation across species. A rice cDNA fragment encoding a putative casein kinase I (CKI) was identified via cDNA macroarray under brassinosteroid (BR) treatment, and a 1939-bp full-length cDNA, OsCKI1, was isolated and
Casein, Dephosphorylated, has been used in studies of modulators of circadian clocks. It has been shown that several molecules can inhibit casein kinase which results in the lenghthening of these clocks.
Chen, Z., et al.
Proceedings of the National Academy of Sciences of the USA (2011)
Marie Lawson et al.
European journal of medicinal chemistry, 123, 105-114 (2016-07-31)
We report here the synthesis, the biological evaluation and the molecular modeling studies of new imidazo[1,2-a]pyridines derivatives designed as potent kinase inhibitors. This collection was obtained from 2-aminopyridines and 2-bromoacetophenone which afforded final compound in only one step. The bioactivity
v-Src-dependent down-regulation of the Ste20-like kinase SLK by casein kinase II
Chaar Z, et al.
Test, 281(38) (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service