Skip to Content
Merck
All Photos(2)

Documents

C3652

Sigma-Aldrich

5α-Cholest-7-en-3β-ol

Synonym(s):

3β-Hydroxy-5α,7-cholestene, Lathosterol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H46O
CAS Number:
Molecular Weight:
386.65
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (TLC)

Quality Level

form

powder

shipped in

ambient

storage temp.

−20°C

SMILES string

CC(CCC[C@@H](C)[C@@](CC[C@]1(C2=CC[C@@]3([H])C[C@@](O)(CC[C@]43C)[H])[H])([C@]1(CC[C@@]24[H])C)[H])C

InChI

1S/C27H46O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h10,18-21,23-25,28H,6-9,11-17H2,1-5H3/t19-,20+,21?,23?,24?,25?,26?,27?/m1/s1

InChI key

IZVFFXVYBHFIHY-IRRCOBOTSA-N

Application

5α-Cholest-7-en-3β-ol was used as a substrate in enzymatic assay for plasma cholesterol. It was used to supplement diet of Daphnia galeata to study the effect on life-history traits.

Biochem/physiol Actions

5α-Cholest-7-en-3β-ol (Lathosterol) is an intermediate in cholesterol biosynthesis pathway. Lathosterol in serum is carried on lipoproteins and is indicative of the rate of cholesterol synthesis. It acts as a marker of synthesis of cholesterol and is not affected by dietary consumption of cholesterol.

Preparation Note

5α-Cholest-7-en-3β-ol yields clear, colorless solution in chloroform at 10 mg/ml.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y H Han et al.
Biological & pharmaceutical bulletin, 23(10), 1247-1249 (2000-10-21)
From the butanol fraction of the starfish Asterina pectinifera Müler et Troschel (Asteriidae), we have isolated a new component, 5alpha-cholest-7-en-3beta-ol. Its antigenotoxic and antimutagenic activities were examined by the SOS chromotest with Escherichia coli PQ37 and by Ames test with
Matthew G K Benesch et al.
Biochemistry, 50(46), 9982-9997 (2011-09-29)
We performed differential scanning calorimetry (DSC) and Fourier transform infrared (FTIR) spectroscopic measurements to study the effects of lathosterol (Lath) on the thermotropic phase behavior and organization of dipalmitoylphosphatidylcholine (DPPC) bilayer membranes and compared our results with those previously reported
Artyom Kopp
Current biology : CB, 22(23), R1004-R1006 (2012-12-12)
Host-plant specialization plays a key role in insect evolution, but little is known about its molecular basis. A new paper shows that a cactus-feeding fly became restricted to its host by changes in an enzyme that converts dietary sterols into
W C Duane
Journal of lipid research, 36(2), 343-348 (1995-02-01)
We measured serum lathosterol levels and whole body cholesterol synthesis by sterol balance in 12 human subjects on a metabolic ward in four randomly allocated, 6-7 week periods: 1) lovastatin (40 mg b.i.d.) + low cholesterol diet (mean 246 mg/day);
A Rahier
Biochemistry, 40(1), 256-267 (2001-01-05)
Deuterium-labeled 5alpha-cholest-7-en-3beta-ol (1) bearing one or two deuteriums at the C-5alpha and (or) C-6alpha positions was synthesized in high isotopic and chiral purity. These compounds were used as substrates with the microsomal wild-type Zea mays and recombinant Arabidopsis thaliana Delta(7)-sterol-C5(6)-desaturases

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service