Skip to Content
Merck
All Photos(5)

Documents

C3130

Sigma-Aldrich

Cortisone 21-acetate

≥99%

Synonym(s):

17α,21-Dihydroxy-4-pregnene-3,11,20-trione 21-acetate, 21-Acetoxy-4-pregnen-17α-ol-3,11,20-trione, 4-Pregnene-17α,21-diol-3,11,20-trione 21-acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H30O6
CAS Number:
Molecular Weight:
402.48
Beilstein:
2067543
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Quality Level

sterility

non-sterile

Assay

≥99%

form

powder

mp

237-240 °C (lit.)

solubility

chloroform: 50 mg/mL, clear, colorless to faintly yellow

shipped in

ambient

storage temp.

room temp

SMILES string

[H][C@@]12CCC3=CC(=O)CC[C@]3(C)[C@@]1([H])C(=O)C[C@@]4(C)[C@@]2([H])CC[C@]4(O)C(=O)COC(C)=O

InChI

1S/C23H30O6/c1-13(24)29-12-19(27)23(28)9-7-17-16-5-4-14-10-15(25)6-8-21(14,2)20(16)18(26)11-22(17,23)3/h10,16-17,20,28H,4-9,11-12H2,1-3H3/t16-,17-,20+,21-,22-,23-/m0/s1

InChI key

ITRJWOMZKQRYTA-RFZYENFJSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Cortisone 21-acetate is a prodrug, which has glucocorticoid and mineralocorticoid properties. It is used as a therapeutic option for glucocorticoid replacement.

Application

Cortisone 21-acetate has been used to study its effects in metastasis and phagocytic activity.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Replacement therapy in Addison?s disease
Lovaas K and Husebye ES
Expert Opinion on Pharmacotherapy (2003)
Immunological mechanisms in metastatic spread and the antimetastatic effects of C. parvum
Jones PD and Castro JE
British Journal of Cancer, 35(5), 519-519 (1977)
Lin Lin et al.
The Journal of infectious diseases, 205(2), 337-347 (2011-11-30)
Activated transfected killer (ATAK) cells are immortal phagocytes transfected with a luminescence reporter that effectively treat lethal infections in neutropenic mice. Their in vivo trafficking, lifespan, and immunogenicity are unknown. Mice were made neutropenic; infected or not with Staphylococcus aureus
Ana Gil-Bona et al.
Journal of proteomics, 127(Pt B), 340-351 (2015-06-19)
The ability to switch from yeast to hyphal growth is essential for virulence in Candida albicans. The cell surface is the initial point of contact between the fungus and the host. In this work, a free-gel proteomic strategy based on
Foye's Principles of Medicinal Chemistry (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service