Skip to Content
Merck
All Photos(2)

Key Documents

C2365

Sigma-Aldrich

Clofilium tosylate

>97%, solid

Synonym(s):

4-Chloro-N,N-diethyl-N-heptylbenzenebutanaminium tosylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H37ClN · C7H7O3S
CAS Number:
Molecular Weight:
510.17
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

>97%

form

solid

color

white

solubility

H2O: soluble

SMILES string

Cc1ccc(cc1)S([O-])(=O)=O.CCCCCCC[N+](CC)(CC)CCCCc2ccc(Cl)cc2

InChI

1S/C21H37ClN.C7H8O3S/c1-4-7-8-9-11-18-23(5-2,6-3)19-12-10-13-20-14-16-21(22)17-15-20;1-6-2-4-7(5-3-6)11(8,9)10/h14-17H,4-13,18-19H2,1-3H3;2-5H,1H3,(H,8,9,10)/q+1;/p-1

InChI key

MOQZYUUHIWPDQC-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

K+ channel blocker; cardiac depressant; anti-arrhythmic

Features and Benefits

This compound is featured on the Potassium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wuyang Wang et al.
The Journal of cell biology, 216(6), 1715-1730 (2017-05-05)
The resting membrane potential (Δψ) of the cell is negative on the cytosolic side and determined primarily by the plasma membrane's selective permeability to K
Clofilium and other class III agents.
Steinberg, et al. et al.
Handbook of Experimental Pharmacology, 89, 389-389 (1989)
H Papp et al.
Journal of physiology and pharmacology : an official journal of the Polish Physiological Society, 67(5), 731-737 (2016-12-25)
Hyperventilation reduces partial pressure of CO
J P Arena et al.
Molecular pharmacology, 34(1), 60-66 (1988-07-01)
The whole-cell arrangement of the patch clamp was used to study delayed rectifier and inward rectifier K channels in isolated guinea pig ventricular cells. Block of these channels by an externally applied quaternary nitrogen compound, clofilium, and two of its
W B Gough et al.
Cardiovascular research, 23(6), 554-559 (1989-06-01)
To determine whether ischaemic tissue is more sensitive than normal tissue to class III anti-arrhythmic agents, we studied the effects of the class III agents clofilium, d-sotalol and bretylium on normal and ischaemic Purkinje fibres one day after ligation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service