Skip to Content
Merck
All Photos(1)

Key Documents

C1999

Sigma-Aldrich

CMP-Sialic Acid Synthetase from Neisseria meningitidis group B

recombinant, expressed in E. coli BL21, ≥10 units/mg protein

Synonym(s):

CTP: N-Acylneuraminate cytidylyltransferase

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

recombinant

expressed in E. coli BL21

Quality Level

form

lyophilized solid

specific activity

≥10 units/mg protein

mol wt

26.0 kDa

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Cytidine monophosphate (CMP)-Sialic Acid Synthetase from Neisseria meningitidis group B is encoded in neuA gene. The protein has a molecular weight of 24.8 kDa.

Application

The enzyme has been utilized to synthesize CMP-sialic acid and its derivatives.

Biochem/physiol Actions

Cytidine monophosphate (CMP)-sialic acid synthetase catalyses the conversion of N?acetylneuraminic acid (NeuNAc) to CMP-NeuNAc. CMP-sialic acid synthetase has globular α/β domain and is categorised under αβα three-layered sandwich fold. The dimerization domain aids the interaction between the monomers. It also has mononucleotide binding and NeuAc binding pocket. Mg2+ is essential for the catalytic functionality of CMP-sialic acid synthetase.

Unit Definition

One unit will catalyze the formation of 1 μmol CMP-Neu-5-Ac from Neu-5-Ac and CTP per minute at 37 °C at pH 8.0.

Physical form

Supplied as a lyophilized powder containing Tris-HCl and NaCl.

Analysis Note

Enzymatic activity assays are performed in Tris-HCl buffer (100 mM, pH 8.5) containing Neu-5-Ac (1 mM) and CTP (1 mM) at 37 °C for 30 min and analyzed using capillary electrophoresis with a UV detector (200 nm).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rahman M Mizanur et al.
Applied microbiology and biotechnology, 76(4), 827-834 (2007-07-03)
In this study, we report the cloning, recombinant expression, and biochemical characterization of a heat-stable CMP-N-acylneuraminic acid (NeuAc) synthetase from Clostridium thermocellum ATCC 27405. A high throughput electrospray ionization mass spectrometry (ESI-MS)-based assay demonstrates that the enzyme has an absolute
Rahman M Mizanur et al.
Applied microbiology and biotechnology, 80(5), 757-765 (2008-08-22)
Sialic acids are abundant nine-carbon sugars expressed terminally on glycoconjugates of eukaryotic cells and are crucial for a variety of cell biological functions such as cell-cell adhesion, intracellular signaling, and in regulation of glycoproteins stability. In bacteria, N-acetylneuraminic acid (Neu5Ac)
Hai Yu et al.
Bioorganic & medicinal chemistry, 12(24), 6427-6435 (2004-11-24)
Three C terminal His6-tagged recombinant microbial CMP-sialic acid synthetases [EC 2.7.7.43] cloned from Neisseria meningitidis group B, Streptococcus agalactiae serotype V, and Escherichia coli K1, respectively, were evaluated for their ability in the synthesis of CMP-sialic acid derivatives in a
Jessica H Wong et al.
Organic & biomolecular chemistry, 7(1), 27-29 (2008-12-17)
A modular replacement approach to the synthesis of sulfo-nucleotide analogs prepared from condensation of nucleoside aldehydes with bis phosphonate Horner-Wadsworth-Emmons reagents is disclosed. These analogs were shown to be inhibitors of Neisseria meningitidis CSS (NmCSS), which is a key enzyme
Sandra Ratzow et al.
Journal of clinical microbiology, 45(6), 1965-1968 (2007-04-06)
The standard sequence-based method for the typing of Legionella pneumophila serogroup 1 strains was extended by using the gspA and neuA alleles. The use of neuA as a seventh allele for typing significantly increased the index of discrimination calculated for

Articles

Explore tools for glycosyltransferase synthesis and modification of glycans, such as glycosyltransferases and nucleotide sugar donors.

Explore tools for glycosyltransferase synthesis and modification of glycans, such as glycosyltransferases and nucleotide sugar donors.

Explore tools for glycosyltransferase synthesis and modification of glycans, such as glycosyltransferases and nucleotide sugar donors.

Explore tools for glycosyltransferase synthesis and modification of glycans, such as glycosyltransferases and nucleotide sugar donors.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service