Skip to Content
Merck
All Photos(2)

Documents

C1832

Sigma-Aldrich

Cyclosporin A

BioReagent, from Tolypocladium inflatum, for molecular biology, ≥95%

Synonym(s):

Antibiotic S 7481F1, Cyclosporine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C62H111N11O12
CAS Number:
Molecular Weight:
1202.61
Beilstein:
3647785
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.52

biological source

Tolypocladium inflatum

Quality Level

grade

for molecular biology

product line

BioReagent

Assay

≥95%

form

powder

technique(s)

drug transporter assay: suitable

solubility

dichloromethane: 9.80-10.20 mg/mL, clear, colorless to faintly yellow

suitability

corresponds to standard
suitable for molecular biology

antibiotic activity spectrum

fungi

Mode of action

enzyme | inhibits

storage temp.

2-8°C

SMILES string

CC[C@@H]1NC(=O)[C@H]([C@H](O)[C@H](C)C\C=C\C)N(C)C(=O)[C@H](C(C)C)N(C)C(=O)[C@H](CC(C)C)N(C)C(=O)[C@H](CC(C)C)N(C)C(=O)[C@@H](C)NC(=O)[C@H](C)NC(=O)[C@H](CC(C)C)N(C)C(=O)[C@@H](NC(=O)[C@H](CC(C)C)N(C)C(=O)CN(C)C1=O)C(C)C

InChI

1S/C62H111N11O12/c1-25-27-28-40(15)52(75)51-56(79)65-43(26-2)58(81)67(18)33-48(74)68(19)44(29-34(3)4)55(78)66-49(38(11)12)61(84)69(20)45(30-35(5)6)54(77)63-41(16)53(76)64-42(17)57(80)70(21)46(31-36(7)8)59(82)71(22)47(32-37(9)10)60(83)72(23)50(39(13)14)62(85)73(51)24/h25,27,34-47,49-52,75H,26,28-33H2,1-24H3,(H,63,77)(H,64,76)(H,65,79)(H,66,78)/b27-25+/t40-,41+,42-,43+,44+,45+,46+,47+,49+,50+,51+,52-/m1/s1

InChI key

PMATZTZNYRCHOR-CGLBZJNRSA-N

Gene Information

human ... PPIA(5478)

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: peptide
Cyclosporin A is a fungal metabolite with immunosuppressive properties. Cyclosporin A is a non-polar cyclic oligopeptide produced by the fungus Tolypocladium inflatum. It is a potent immunosuppressive agent, affecting primarily T-lymphocytes.

Application

Cyclosporin A was suitable for:

  • Using in in vivo Neovascularization Assay, to subcutaneously inject the mice daily for suppressing the immune response.
  • Measuring the multiple drug resistance transporter activity in putative cancer stem/progenitor cells.
  • mRNA transcription studies
  • analytical standard

Biochem/physiol Actions

A fungal metabolite possessing potent immunosuppressive properties. It inhibits the T-cell receptor signal transduction pathway via the formation of cyclosporin A−cyclophilin complex that inhibits calcineurin (protein phosphatase 2B). Inhibits nitric oxide synthesis induced by interleukin 1α, lipopolysaccharides and TNFα. Can block cytochrome c release from mitochondria.
Cyclosporin A has been shown to inhibit the functioning of several nuclear proteins involved in T-cell activation at the level of mRNA transcription. It forms a complex with its intracellular receptor cyclophilin, which can then bind to calcineurin, a Ca2+ - and calmodulin-dependent protein phosphatase, inhibiting its enzymatic activity. It was found to suppress the replication of hepatitis C virus genome in cultured hepatocytes.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R A Nichols et al.
The Journal of biological chemistry, 269(38), 23817-23823 (1994-09-23)
In response to Ca2+ entry, several prominent brain nerve terminal phosphoproteins undergo dephosphorylation, but the relation between dephosphorylation and neurotransmitter release is unknown. Using the immunosuppressants cyclosporin A (CsA) and L-683,590 (FK-520) to inhibit specifically the Ca2+/calmodulin-dependent protein phosphatase calcineurin
Mi-Ok Lee et al.
Arteriosclerosis, thrombosis, and vascular biology, 32(2), 343-352 (2011-11-15)
A number of studies have revealed that stress signaling and subsequent stress responses in stem/progenitor cells are responsible for attenuated regeneration or degenerative disease. Because ionizing radiation (IR), which sensitizes diverse types of stem cells, reportedly induces cardio-circulatory diseases, we
Koichi Watashi et al.
Hepatology (Baltimore, Md.), 38(5), 1282-1288 (2003-10-28)
Persistent infection of hepatitis C virus (HCV) is a major cause of liver diseases such as chronic hepatitis, liver cirrhosis, and hepatocellular carcinoma. Searching for a substance with anti-HCV potential, we examined the effects of a variety of compounds on
N A Clipstone et al.
The Journal of biological chemistry, 269(42), 26431-26437 (1994-10-21)
The calcium/calmodulin-regulated phosphatase calcineurin (CN) is the site of action of the immunosuppressive drugs cyclosporin A (CsA) and FK506. CN has recently been established as a key signaling enzyme in the T cell signal transduction cascade and an important regulator
Mohammed E Abo-El Fetoh et al.
Naunyn-Schmiedeberg's archives of pharmacology, 393(5), 897-908 (2020-01-08)
One of the most common causes of cancer mortality worldwide is hepatocellular carcinoma (HCC). Extracellular signal-regulated kinase (ERK1/2) pathway has been shown to play an important role in the development and progression of HCC. Here, we demonstrate that the immunosuppressive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service