Skip to Content
Merck
All Photos(1)

Key Documents

PHR1092

Supelco

Cyclosporine

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Cyclosporin A, Antibiotic S 7481F1, Cyclosporine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C62H111N11O12
CAS Number:
Molecular Weight:
1202.61
Beilstein:
3647785
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. C2163000
traceable to USP 1158504

API family

cyclosporine

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

-10 to -25°C

SMILES string

CC[C@@H]1NC(=O)[C@H]([C@H](O)[C@H](C)C\C=C\C)N(C)C(=O)[C@H](C(C)C)N(C)C(=O)[C@H](CC(C)C)N(C)C(=O)[C@H](CC(C)C)N(C)C(=O)[C@@H](C)NC(=O)[C@H](C)NC(=O)[C@H](CC(C)C)N(C)C(=O)[C@@H](NC(=O)[C@H](CC(C)C)N(C)C(=O)CN(C)C1=O)C(C)C

InChI

1S/C62H111N11O12/c1-25-27-28-40(15)52(75)51-56(79)65-43(26-2)58(81)67(18)33-48(74)68(19)44(29-34(3)4)55(78)66-49(38(11)12)61(84)69(20)45(30-35(5)6)54(77)63-41(16)53(76)64-42(17)57(80)70(21)46(31-36(7)8)59(82)71(22)47(32-37(9)10)60(83)72(23)50(39(13)14)62(85)73(51)24/h25,27,34-47,49-52,75H,26,28-33H2,1-24H3,(H,63,77)(H,64,76)(H,65,79)(H,66,78)/b27-25+/t40-,41+,42-,43+,44+,45+,46+,47+,49+,50+,51+,52-/m1/s1

InChI key

PMATZTZNYRCHOR-CGLBZJNRSA-N

Gene Information

human ... PPIA(5478)

Looking for similar products? Visit Product Comparison Guide

General description

Cyclosporine is a lipophilic immunosuppressive peptide with antifungal and anti-inflammatory properties. It selectively inhibits the production of cytokines involved in T-lymphocyte activation in response to antigen stimulation. It is widely used to prevent transplanted organ rejection and for the treatment of various autoimmune diseases.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.
Cyclosporine may be used as a reference standard for the determination of cyclosporine in biological samples by high-performance liquid chromatography tandem mass spectrometry (HPLC-MS/MS) method.

Biochem/physiol Actions

A fungal metabolite possessing potent immunosuppressive properties. It inhibits the T-cell receptor signal transduction pathway via the formation of cyclosporin A−cyclophilin complex that inhibits calcineurin (protein phosphatase 2B). Inhibits nitric oxide synthesis induced by interleukin 1α, lipopolysaccharides and TNFα. Can block cytochrome c release from mitochondria.
Potent immunosuppressant; inhibits nitric oxide synthesis induced by interleukin 1α, lipopolysaccharides and TNFα; blocks cytochrome c release from mitochondria.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB3757 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tip W Loo et al.
Biochemistry, 52(41), 7167-7169 (2013-10-03)
There is no high-resolution crystal structure of the human P-glycoprotein (P-gp) drug pump. Homology models of human P-gp based on the crystal structures of mouse or Caenorhabditis elegans P-gps show large differences in the orientation of transmembrane segment 5 (TM5).
Jane Rasaiyaah et al.
Nature, 503(7476), 402-405 (2013-11-08)
Human immunodeficiency virus (HIV)-1 is able to replicate in primary human macrophages without stimulating innate immunity despite reverse transcription of genomic RNA into double-stranded DNA, an activity that might be expected to trigger innate pattern recognition receptors. We reasoned that
Koichi Watashi et al.
Hepatology (Baltimore, Md.), 59(5), 1726-1737 (2014-01-01)
Chronic hepatitis B virus (HBV) infection is a major public health problem worldwide. Although nucleos(t)ide analogs inhibiting viral reverse transcriptase are clinically available as anti-HBV agents, emergence of drug-resistant viruses highlights the need for new anti-HBV agents interfering with other
David A Williams et al.
Pediatric blood & cancer, 61(5), 869-874 (2013-11-29)
Randomized clinical trials in pediatric aplastic anemia (AA) are rare and data to guide standards of care are scarce. Eighteen pediatric institutions formed the North American Pediatric Aplastic Anemia Consortium to foster collaborative studies in AA. The initial goal of
Hae-Rim Kim et al.
Arthritis & rheumatology (Hoboken, N.J.), 66(3), 538-548 (2014-02-28)
Stromal cell-derived factor 1 (SDF-1) is a chemokine that is involved in the bone-destructive process in rheumatoid arthritis (RA) and bony metastasis in malignancy. This study was undertaken to determine the role and mechanism of SDF-1 in RA-associated osteoclastogenesis. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service