Skip to Content
Merck
All Photos(1)

Documents

C1145

Sigma-Aldrich

Cholesterol–PEG 600

Synonym(s):

Cholesterol, water soluble from sheep, Cholesteryl-polyethylene glycol 600 sebacate, Polyethylene glycol 600 mono(cholesteryl) ether sebacate, Polyoxyethyl-cholesteryl sebacate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41141804
eCl@ss:
39023139
NACRES:
NA.77

form

solid

extent of labeling

≥20% cholesterol

color

light yellow

solubility

H2O: 60 mg/mL

shipped in

dry ice

storage temp.

−20°C

General description

Cholesterol is one of the important constituent of all membranes. It is made up of a hydrophobic, planar, a polar hydroxyl group, fused-ring nucleus and a short hydrocarbon tail.

Application

Cholesterol-PEG 600 has been used to evaluate its ability to induce apoptosis in vascular smooth muscle cells. It has also been used to restore cholesterol following depletion by methyl β-cyclodextrin.

Biochem/physiol Actions

Cholesterol acts as an antecedent of steroid hormones. It plays a key role in fetal growth and in brain development.

Physical form

semisolid to solid

Preparation Note

Prepared by method of Proksch, G.J., and Bonderman, D.P., Clin. Chem., 24, 1924 (1978).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Absi et al.
British journal of pharmacology, 151(3), 332-340 (2007-04-24)
The small and intermediate conductance, Ca2+-sensitive K+ channels (SK(Ca) and IK(Ca), respectively) which are pivotal in the EDHF pathway may be differentially activated. The importance of caveolae in the functioning of IK(Ca) and SK(Ca) channels was investigated. The effect of
The Lipids of Biological Membranes
The Membranes of Cells null
Linda Shaw et al.
Cardiovascular research, 69(4), 825-835 (2006-02-09)
A close association of caveolae and sarcoplasmic reticulum (SR) has been suggested to be important for contractile activation of smooth muscle. Here, we investigate the presence of such arrangements in pressurised resistance arteries and examine the influence of two agents
Apoptosis of vascular smooth muscle cells induced by cholesterol and its oxides in vitro and in vivo
Yin J, et al.
Atherosclerosis, 148(2), 365-374 (2000)
Disruption of cholesterol homeostasis in developmental neurotoxicity
Reproductive and Developmental Toxicology null

Articles

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service