Skip to Content
Merck
All Photos(1)

Key Documents

700143P

Avanti

27-alkyne cholesterol

Avanti Research - A Croda Brand 700143P, powder

Synonym(s):

27-alkyne cholesterol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H44O
CAS Number:
Molecular Weight:
396.65
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (700143P-1MG)
pkg of 1 × 100 μg (700143P-100ug)
pkg of 1 × 250 μg (700143P-250ug)

manufacturer/tradename

Avanti Research - A Croda Brand 700143P

shipped in

dry ice

storage temp.

−20°C

General description

Alkyne cholesterol is used to trace cholesterol metabolism and localization.
This reagent is a modified lipid containing an omega-terminal alkyne. The terminal alkyne group can be used in a highly specific linking reaction with azide-containing reagents, known as ‘click chemistry′, in the presence of a copper (Cu)-containing catalyst.

Biochem/physiol Actions

27-alkyne cholesterol acts similar to natural sterols such as, ergosterol and cholesterol.

Packaging

5 mL Amber Glass Screw Cap Vial (700143P-1MG)
5 mL Amber Glass Screw Cap Vial (700143P-100ug)
5 mL Amber Glass Screw Cap Vial (700143P-250ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A novel alkyne cholesterol to trace cellular cholesterol metabolism and localization.
Hofmann K, et al.
Journal of Lipid Research, 55(3), 583-591 (2014)
Yaroslav V Faletrov et al.
Chemistry and physics of lipids, 227, 104850-104850 (2019-12-15)
20-hydroxycholesterol is a signaling oxysterol with immunomodulating functions and, thus, structural analogues with reporter capabilities could be useful for studying and modulating the cellular processes concerned. We have synthesized three new 20-hydroxycholesterol-like pregn-5-en-3β-ol derivatives with fluorescent 7-nitrobenzofurazan (NBD) or Raman-sensitive
Kristina Hofmann et al.
Journal of lipid research, 55(3), 583-591 (2013-12-18)
Cholesterol is an important lipid of mammalian cells and plays a fundamental role in many biological processes. Its concentration in the various cellular membranes differs and is tightly regulated. Here, we present a novel alkyne cholesterol analog suitable for tracing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service