Skip to Content
Merck
All Photos(1)

Key Documents

C0733

Sigma-Aldrich

Cyclooxygenase 1 from sheep

glycerol solution, ≥1500 units/mg protein

Synonym(s):

COX-1, Constitutive cyclooxygenase, Prostaglandin H synthase 1, Prostaglandin endoperoxide synthase

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.32

form

glycerol solution

Quality Level

specific activity

≥1500 units/mg protein

mol wt

dimer subunit mol wt 70 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

sheep ... COX1(808251)

General description

Cyclooxygenase 1 is a 71 kDa membrane bound protein predominantly present in endoplasmic reticulum. It has three domains, the epidermal growth factor (EGF) like domain, enzymatic and membrane binding domain. Cyclooxygenase 1 mediates prostaglandin synthesis and is modulated by anti inflammatory nonsteroidal drugs.

Application

Cyclooxygenase 1 from sheep has been used as positive control protein in western blot analysis of osteoarthritis samples and in prostaglandin synthase activity assay.

Biochem/physiol Actions

COX-1 catalyzes the conversion of arachidonic acid to prostaglandin H2 (the first step in the biosynthesis of prostaglandins, thromboxanes, and prostacyclins). It is involved in the homeostatic role of eicosanoids and constitutively expressed in almost all animal tissues. Has an apparent KM of 8.3 μM for arachidonic acid.

Unit Definition

One unit consumes one nanomole of oxygen per minute at 37 °C in 0.1 M Tris-HCl buffer, pH 8, containing 100 μM arachidonate, 5 mM EDTA, 2 mM phenol, and 1 μM hematin.

Physical form

Solution in 80 mM Tris-HCl, pH 8, with 0.1% TWEEN® 20, 300 μM diethyldithiocarbamate and 10% glycerol.

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cyclo-oxygenase isoenzymes: physiological and pharmacological role
Kam PCA and See AUL
Anaesthesia, 55(5), 442-449 (2000)
Maurizio Anzini et al.
Journal of medicinal chemistry, 56(8), 3191-3206 (2013-03-29)
A series of 3-substituted 1,5-diarylpyrroles bearing a nitrooxyalkyl side chain linked to different spacers were designed. New classes of pyrrole-derived nitrooxyalkyl inverse esters, carbonates, and ethers (7-10) as COX-2 selective inhibitors and NO donors were synthesized and are herein reported.
S Ohki et al.
The Journal of biological chemistry, 254(3), 829-836 (1979-02-10)
The highly purified prostaglandin endoperoxide synthetase from bovine vesicular gland microsomes had two still unresolved enzyme activities; the oxygenative cyclization of 8,11,14-eicosatrienoic acid to produce prostaglandin G1 and the conversion of the 15-hydro-peroxide of prostaglandin G1 to a 15-hydroxyl group
The Sigma class glutathione transferase from the liver fluke Fasciola hepatica
LaCourse EJ, et al.
PLoS Neglected Tropical Diseases, 6(5), e1666-e1666 (2012)
Expression and activity of COX-1 and 2 and 5-LOX in joint tissues from dogs with naturally occurring coxofemoral joint osteoarthritis
Lascelles BD, et al.
Journal of Orthopaedic Research : Official Publication of the Orthopaedic Research Society, 27(9), 1204-1208 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service