Skip to Content
Merck
All Photos(3)

Key Documents

128708

Sigma-Aldrich

trans-Ferulic acid

99%

Synonym(s):

trans-4-Hydroxy-3-methoxycinnamic acid, Ferulic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H3(OCH3)CH=CHCO2H
CAS Number:
Molecular Weight:
194.18
Beilstein:
1570363
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

powder
powder and chunks

mp

168-172 °C (lit.)

functional group

carboxylic acid

SMILES string

OC(/C=C/C1=CC(OC)=C(O)C=C1)=O

InChI

1S/C10H10O4/c1-14-9-6-7(2-4-8(9)11)3-5-10(12)13/h2-6,11H,1H3,(H,12,13)/b5-3+

InChI key

KSEBMYQBYZTDHS-HWKANZROSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

trans-Ferulic acid was used for esterification of methacrylated dextran. It was also used to study the effect of oral administration of trans-ferulic acid to c57BL mice on ethanol-induced liver injury. Furthermore, it has been used in the synthesis of new biocompatible antioxidant polymers linking trans-ferulic acid.

Biochem/physiol Actions

Hydroxycinnamic acid found in plant-based foods. Antioxidant.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roberta Cassano et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 72(1), 232-238 (2008-11-04)
Long-term exposure of the skin to UV light causes degenerative effects, which can be minimized by using antioxidant formulations. The major challenge in this regard is that a significant amount of antioxidant should reach at the site for effective photoprotection.
Chatchawan Chotimarkorn et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 15(11), 951-958 (2008-04-22)
The effects of the oral administration of trans-ferulic acid and gamma-oryzanol (mixture of steryl ferulates) with ethanol (5.0 g per kg) for 30 days to c57BL mice on ethanol-induced liver injury were investigated. Preventions of ethanol-induced liver injury by trans-ferulic
Helle Harder et al.
European journal of nutrition, 43(4), 230-236 (2004-08-17)
Rye bread contributes an important part of the whole grain intake in the Scandinavian diet. Ferulic acid is the major phenolic compound in rye bran and is an antioxidant in vitro and may, therefore, contribute to cardioprotective effects of whole
Sonia Trombino et al.
Journal of agricultural and food chemistry, 52(8), 2411-2420 (2004-04-15)
Although an antioxidant mechanism has been involved in the beneficial effects of ferulic acid in human diseases, there are few reports on the antioxidant properties of this compound in isolated membranes and intact cells. Here, we evaluated the ability of
Nickolas A Anderson et al.
The Plant cell, 27(8), 2195-2209 (2015-08-13)
Modifying lignin composition and structure is a key strategy to increase plant cell wall digestibility for biofuel production. Disruption of the genes encoding both cinnamyl alcohol dehydrogenases (CADs), including CADC and CADD, in Arabidopsis thaliana results in the atypical incorporation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service