Skip to Content
Merck
All Photos(2)

Documents

C4049

Sigma-Aldrich

Chlorocholine chloride

suitable for plant cell culture, BioReagent

Synonym(s):

Cycocel, (2-Chloroethyl)trimethylammonium chloride, Chlormequat chloride, Choline dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3N(Cl)CH2CH2Cl
CAS Number:
Molecular Weight:
158.07
Beilstein:
3563994
EC Number:
MDL number:
UNSPSC Code:
10171502
PubChem Substance ID:
NACRES:
NA.72

product line

BioReagent

form

powder

technique(s)

cell culture | plant: suitable

application(s)

agriculture

SMILES string

[Cl-].C[N+](C)(C)CCCl

InChI

1S/C5H13ClN.ClH/c1-7(2,3)5-4-6;/h4-5H2,1-3H3;1H/q+1;/p-1

InChI key

UHZZMRAGKVHANO-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Chlorocholine chloride/Chlormequat Chloride (CCC) is a plant growth regulator. It is an irritant and a skin absorbant.

Biochem/physiol Actions

Chlormequat (Chlorocholine) chloride is a synthetic plant growth regulator that inhibits gibberellin (GA) biosynthesis. It is used to study the mechanisms and effects of gibberellins on plant cell growth and differentiation.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nadjimov et al.
Journal of plant growth regulation, 18(1), 45-48 (1999-11-30)
The round-leafed mutant cotton line L-501 developed fasciation of the upper stem when field grown in Central Asia. Fasciation co-segregated with the mutant gene for round leaves In.(l) Fasciation developed at the flowering stage, but removal of floral buds did
J D Metzger
Plant physiology, 94(1), 151-156 (1990-09-01)
Field pennycress (Thlaspi arvense L.) is a winter annual weed with a cold requirement for stem elongation and flowering. The relative abilities of several native gibberellins (GAs) and GA-precursors to elicit stem growth were compared. Of the eight compounds tested
C
Sittig's Handbook of Pesticides and Agricultural Chemicals, 91-195 (2015)
J D Metzger
Plant physiology, 86(1), 237-240 (1988-01-01)
Petiole growth in Thlaspi arvense L. was stimulated when a basic 8 hour photoperiod (4.20 milliwatts per square centimeter) was extended with low intensity light (0.16 milliwatt per square centimeter) from incandescent lamps. The day length extension was effective only
René K Juhler et al.
Environmental science and pollution research international, 17(6), 1245-1256 (2010-02-24)
Chlormequat (Cq) is a plant growth regulator used throughout the world. Despite indications of possible effects of Cq on mammalian health and fertility, little is known about its fate and transport in subsurface environments. The aim of this study was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service